Skip to Content
Merck
All Photos(1)

Key Documents

SCT026

Sigma-Aldrich

BioTracker 510 Green β-Glu Dye

Live cell imaging dye for Beta-glucosidase (β-Glu).

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.47

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Beta-glucosidase (β-Glu) catalyzes the hydrolysis of the glycosidic bonds to terminal non-reducing residues in beta-D-glucosides and oligosaccharides, with release of glucose.

The BioTracker 510 Green β-Glu Dye is permeable through the cell membrane and is fluorescent substrate [9- (4′-methoxy-2′-methylphenyl) -6- (β-D-glucopylanosyloxy) -xanthen-3-one] for detecting β-glucosidase in living cells. Non-fluorescent dye is hydrolyzed by the β-Glucosidase, and generates bright green fluorescence (510 nm) when it is irradiated by the 490 nm excitation light.

Spectral Properties
Absorbance: 492nm
Emission: 510nm

Application

Live cell fluorescent imaging
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Physical form

Lyophilized

Storage and Stability

Store BioTracker 510 Green β-Glu Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

176.0 °F - (Dimethylsulfoxide)

Flash Point(C)

80 °C - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service