Skip to Content
Merck
All Photos(1)

Key Documents

SCR055

Sigma-Aldrich

ENStem Human Neural Progenitor Expansion Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
Pricing and availability is not currently available.

description

Cell line type: Neural Stem Cells
Neural Stem Cells

Quality Level

technique(s)

cell culture | stem cell: suitable

General description

The ENStem-A Human Neural progenitor Expansion Kit contains ENStem-A Human Neural Progenitors and optimized Expansion medium. ENStem-A Human Neural Progenitor Cells are derived from NIH approved H9 human embryonic stem cells (hESCs). Using a proprietary method developed by Aruna Biomedical, these hESC derived neural progenitors proliferate as an adherent cell monolayer and can readily differentiate into different neuronal subtypes. Detailed spectral analyses indicate ENStem-A cells retained a normal diploid karyotype in culture after ten passages. ENStem-ATM human neural progenitor cells may be used for a variety of research applications such as studies of neurotoxicity, neurogenesis, electrophysiology, neurotransmitter and receptor functions. ENStem-A Neural Expansion Medium is a defined serum-free formulation that has been optimized for the culture and expansion of ENStem-A Human Neural Progenitor Cells. When used in conjunction with FGF-2 (provided), the expansion medium will allow for the maintenance and proliferation of ENStem-A Human Neural Progenitor Cells.

Components

ENSTem-A Human Neural Progenitors, 1 x 106 cells ENStem-A Neural Expansion Media, SCM004, 500 mls Basic FGF, lyophilized, 10 ug

Quality

Each lot of ENStem-A Human Neural Progenitor Cells has been validated for high levels of expression of Nestin and Sox 2, and low level expression of Oct-4. The ability of ENStem-A cells to differentiate into multiple neuronal phenotypes and maintain a normal karyotype after multiple passages has been verified (please refer to datasheet figures). The cells have been confirmed to be negative for mycoplasma.

Storage and Stability

EnStem Human Neural Progenitor Cells: When stored at the recommended storage conditions (liquid nitrogen), ENStem-A cells are stable up to five years. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date. We recommend that the cells be used within ten passages. ENStem-A Human Neural Expansion media ENStem-A Neural Expansion Medium should be stored at -20C until ready to use. Upon thawing, the expansion medium should be stored at 2-8C and given a 1-month expiration dating. Dispense into aliquots to avoid repeated heating prior to each use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service