Skip to Content
Merck
All Photos(1)

Documents

SCM307

Sigma-Aldrich

3dGRO® Lung Organoid Branching Medium

Serum-free ready-to-use media used to differentiate anterior foregut endoderm (AFE) cells into lung bud organoids in suspension cultures.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

Related Categories

General description

The lung epithelium undergoes a complex series of endoderm–mesoderm-mediated signaling events to generate a complex arborized network of conducting airway (bronchi, bronchioles) and gas-exchanging units (alveoli). Lung epithelium differentiation stages include: anterior/posterior and dorsal/ventral patterning, lung specification, lung budding, branching morphogenesis, and maturation.
The 3dGRO® Lung Organoid Branching Medium is a chemically defined/serum-free ready-to-use media used to differentiate Sox2+/Pax6+ anterior foregut endoderm (AFE) cells into lung bud organoids in suspension cultures. Additionally, the 3dGRO® Lung Organoid Branching Medium can be used to culture AFE cells on fibronectin coated cultureware. The 3dGRO® Lung Organoid Branching Medium is a component of the 3dGRO® Human Lung Organoid Culture System which can be used to differentiate human iPS cells into mature lung organoids that structurally resemble the in vivo branching airway and early alveolar structures. The 3dGRO® Human Lung Organoid Culture System includes (all sold separately):

  • Definitive Endoderm Induction Medium (SCM302)
  • Anterior Foregut Endoderm Induction Medium I (SCM305)
  • Anterior Foregut Endoderm Induction Medium II (SCM306)
  • 3dGRO® Lung Organoid Branching Medium (SCM307)
  • 3dGRO® Lung Organoid Maturation Medium (SCM308)

Human iPSC Derived AFE Progenitors (SCC301)

Storage and Stability

Upon receipt, store at -20°C. When ready to use, thaw overnight at 2-8°C. Once thawed, mix thoroughly. Use immediately and store at 2-8°C for up to 1 week. Do not re-freeze. Unused aliquots may be stored at -20°C until the expiry date.

Legal Information

3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Organoid culture FAQs. Learn how to culture and analyze organoids following established 3D cell culture protocols. Tips and tricks.

Related Content

Explore our portfolio of 3D cell culture tools and technologies for any 3D cell line or application and discover organoids, hydrogels, ULA, microwell, or hydrogel plates, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service