Skip to Content
Merck
All Photos(1)

Key Documents

SCM132

Sigma-Aldrich

PluriSTEM-XF® Human ES/iPS Cell Medium

PluriSTEM-XF Human ES/iPS Medium is a complete xeno-free medium formulation for the feeder-free culture of human ES and iPS cells.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

PluriSTEM®

technique(s)

cell culture | embryo: suitable
cell culture | stem cell: suitable

shipped in

dry ice

General description

PluriSTEM-XF Human ES/iPS Medium is a complete xeno-free medium formulation for the feeder-free culture of human ES and iPS cells. Pluripotent cells maintained in thissmall molecule based medium exhibit high cell health, express high levels of pluripotency markers (NANOG, OCT3/4, SOX-2, SSEA-3, SSEA-4, TRA-1-60, and TRA-1-81), retain differentiation potential and possess a normal karyotype. PluriSTEM-XF Human ES/iPS Medium is provided as a stand-alone 500 mL bottle that is ready-to-use and does not require additional supplementation.

For a complete xeno-free culture system, PluriSTEM-XF Human ES/iPS Medium should be used with PluriSTEM-XF Recombinant Vitronectin (Cat. No. CC130).

Each lot of PluriSTEM-XF Human ES/iPS Medium is rigorously quality control tested for the ability to sustain undifferentiated human ES/iPS cells for 3 passages.

Note: Before starting PluriSTEM-XF, users are recommended to read the manual carefully and completely. Colony morphology and growth rates may differ from other serum-free, feeder-free media culture conditions.

Application

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Each lot of PluriSTEM® Human ES/iPS Medium is rigorously quality control tested for the ability to sustain undifferentiated human ES cells for greater than 3 passages.

Storage and Stability

PluriSTEM-XF Human ES/iPS Medium is provided frozen and can be stored at -20°C until the expiration date on the product label.

Before use, thaw PluriSTEM-XF medium at room temperature (15 – 25°C) or overnight at 2 – 8°C. Do not thaw at 37°C. Upon thawing, PluriSTEM-XF medium may be aseptically dispensed into working aliquots and stored at -20°C until the expiration date as indicated on the label. Thawed aliquots may be stored at 2 – 8°C for up to 2 weeks. Do not refreeze aliquots after thawing. Before use, warm working aliquots to room temperature. Do not warm the medium in a 37°C water bath.

Legal Information

PLURISTEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Skip weekend feedings. Defined serum-free and feeder-free expansion media for human pluripotent stem cells (ES and iPS cells). See publications and protocols.

Fibroblast growth factors (FGFs) regulate developmental pathways and mesoderm/ectoderm patterning in early embryonic development.

The Simplicon™ RNA Reprogramming Technology is a next generation reprogramming system that uses a single synthetic, polycistronic self-replicating RNA strand engineered to mimic cellular RNA to generate human iPS cells.

Protocols

Step-by-step stem cell culture protocols for human induced pluripotent stem cells (iPSCs) including ips cell thawing, expanding, freezing and characterizing.

Stem cell reprogramming protocols to generate human induced pluripotent stem cells (iPSCs) including viral and non-viral RNA based methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service