SCC415
BG-1 Human Ovarian Adenocarcinoma Cell Line
Human
Synonym(s):
BG-1 Human Ovarian Adenocarcinoma Cell Line
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
BG-1 Human Ovarian Adenocarcinoma Cell Line,
biological source
human
packaging
pkg of >1X10⁶ vials
manufacturer/tradename
Sigma-Aldrich
growth mode
N/A
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
≤ − 140°C
General description
The BG-1 human ovarian adenocarcinoma cell line is one of the most widely used cell lines in ovarian cancer research, characterized by clinically relevant expression levels of both progesterone receptor and estrogen receptor and a highly robust response to exogenously-applied estrogen in vitro.2 BG-1 cells secrete cancer antigen 125 (CA-125)3 and expressed E-cadherin, an epithelial cell marker.4 The BG-1 human ovarian adenocarcinoma was derived from a stage III primary solid tumor and is a representative model of the most common ovarian cancers.
Application
Each vial contains ≥1X10⁶ cells/vial viable cells.Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, Chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services Cells are negative for mycoplasma contamination.
Worldwide, ovarian cancer is estimated to cause over 140,000 deaths annually.1 The difficulty of early detection means that nearly 3 out of 4 cases are diagnosed at an advanced stage,1 highlighting the necessity of clinically relevant cellular models for understanding the ovarian cancer physiology at advanced stages. Ovarian cancers retaining functional expression of progesterone receptor and estrogen receptors typically have a better prognosis than cancers lacking expression of these receptors, making these carcinomas amenable to therapeutic intervention.The BG-1 human ovarian adenocarcinoma cell line is one of the most widely used cell lines in ovarian cancer research, characterized by clinically relevant expression levels of both progesterone receptor and estrogen receptor and a highly robust response to exogenously-applied estrogen in vitro.2 BG-1 cells secrete cancer antigen 125 (CA-125)3 and expressed E-cadherin, an epithelial cell marker.4 The BG-1 human ovarian adenocarcinoma was derived from a stage III primary solid tumor and is a representative model of the most common ovarian cancers.Source The BG-1 cell line was derived from a poorly differentiated stage III solid primary ovarian tumor from a patient of unspecified age.2 SCC415 has been genetically validated as an authentic BG-1 cell line.References1. Jemal A, Bray F, Center MM, FerlayJ, Ward E, Forman D. CA Cancer J Clin. 2011; 61(2):69-90.2. Geisinger KR et al. Cancer. 1989; 63(2):280-288.3. Li Y et al. Mol Endocrinol 2014; 28(12):2072-2081. 4. Yi BR, Kim TH, Kim YS, Choi KC. Int J Oncol 2015; 46(1): 272-280.
Features and Benefits
BG-1 cancer cell line is a good model for ovarian adenocarinoma
Storage and Stability
The cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.
Other Notes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service