Skip to Content
Merck
All Photos(1)

Documents

RW0629325

Millipore

Reinforced Membrane, mixed cellulose esters, 0.5 µm, cellulose web 293 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602

material

mixed cellulose esters (MCE)
plain filter
web cellulose
white filter

Quality Level

feature

hydrophilic

parameter

550 °C max. temp.
96 mL/min-cm2 water flow rate

filter diam.

293 mm

thickness

140 μm

pore size

0.5 μm pore size
75 % porosity

General description

Filter Code: RW06
Filter Type: Screen filter

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service