Skip to Content
Merck
All Photos(3)

Key Documents

RDYSTRM40

Millipore

ReadyStream® Bag Holder Large

For use with the ReadyStream® system to hold larger size sample bags on the dispensing unit

Synonym(s):

Media Bag Support, Sample bag holder, Stomacher bag holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13102014
NACRES:
NB.24
Pricing and availability is not currently available.

manufacturer/tradename

ReadyStream®

application(s)

food and beverages

compatibility

for use with ReadyStream® system

General description

ReadyStream® Large Bag Holder is an integral part of ReadyStream® Media Dispensing Unit.
It is designed to support the media bag during the dispensing process and is an integral part of the ReadyStream"] Media Dispensing Unit.

Application

ReadyStream® Large Bag Holder is used to support and protect the sample bag during the dispensing process.

Other Notes

Components required but not included: ·

This catalog number doesn’t include all components that are required for the installation of the entire ReadyStream® System.

The following parts need to be ordered separately along with this product:
  • RDYSTRM10.
  • RDYSTRM20.
  • RDYSTRM30.
  • RDYSTRM90.
  • RDYSTRAIN01.


A country specific Power Cord adapted to local electrical requirements. ·

The indicated Price currently corresponds to the above catalog number only.

To access the ReadyStream® Data Matrix Generator Tool follow this link https://www.sigmaaldrich.com/US/en/support/calculators-and-apps/readystream-data-matrix-generator

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Salmonella pass through the entire food chain from animal feed, primary production, and into households or food-service establishments.

Related Content

The Revolution in Media Preparation - New solution for media preparation and dispensing for food testing labs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service