QTUM0TIX1
Quantum® Polishing Cartridge
For Milli-Q® Advantage A10 / Integral / Reference systems connected to ultrapure water feed
Synonym(s):
Quantum® TIX Polishing Cartridge
About This Item
Recommended Products
Quality Level
feature
Removes trace levels of organics and ions from ultrapure water
packaging
pkg of 1 unit
manufacturer/tradename
Quantum®
resistivity
>18.0 MΩ-cm (at 25 degree celcius)
total organic carbon (TOC) residue
<10 ppb
capacity
40 g ion exchange capacity (NaCl)(Dynamic Capcity is greater than 20g NaCl)
compatibility
for use with Milli-Q® Advantage A10
for use with Milli-Q® Integral
for use with Milli-Q® Reference
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Genuine Milli-Q® consumable for the replacement of QTUM0TIX1 - Quantum® TIX Polishing Cartridge.
- Contains Ionex resins for trace ion removal.
- Care-free maintenance: easily installed and replaced.
- e-Sure tag for RFID connection enables full traceability (data management).
- Uses ion exchange technology for purification.
- Certificate of Quality available online.
Components
- Organex (Mix of Synthetic activated carbon & virgin Ion exchange resins)
- Virgin Ion Exchange Resins
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service