Skip to Content
Merck
All Photos(1)

Key Documents

PYYT-66HK

Sigma-Aldrich

Human PYY (Total) RIA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84
Pricing and availability is not currently available.

grade

for protein analysis

Quality Level

description

ED<sub>80</sub> = 37.2 ± 8 pg/mL ED<sub>50</sub> = 110.9 ± 20 pg/mL ED<sub>20</sub> = 333.8 ± 73 pg/mL
analytes available: PYY
10-1280 pg/mL (standard curve range)

feature

accuracy 83–111%

species reactivity

human

sensitivity measuring range

10 pg/mL

technique(s)

radioimmunoassay: suitable

UniProt accession no.

detection method

radioactive

General description

FUNCTION: SwissProt: P10082 # This gut peptide inhibits exocrine pancreatic secretion, has a vasoconstrictory action and inhibitis jejunal and colonic mobility.
SIZE: 97 amino acids; 11046 Da
SUBCELLULAR LOCATION: Secreted.
SIMILARITY: SwissProt: P10082 ## Belongs to the NPY family.

Application

Two-day disequilibrium assay, incubation at 4°C Sample volume: 100 μL serum, plasma, or tissue culture sample

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service