Skip to Content
Merck
All Photos(1)

Key Documents

PX0396D

Supelco

Perchloric Acid 60%

meets analytical specifications of USP/NF GR ACS

Synonym(s):

Perchloric acid, Perchloric Acid, PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352301
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

meets analytical specifications of USP/NF GR ACS

form

liquid

concentration

60.0-62.0% HClO4

impurities

≤0.001% Chloride (Cl)
≤0.001% Nitrogen compounds (as N)
≤0.001% Sulfate (SO4)
≤1 ppm Heavy metals (as Pb)
≤1 ppm Iron (Fe)
≤5 ppm Silicate and phosphate (as SiO2)

ign. residue

≤0.003%

color

APHA: ≤10

shipped in

ambient

storage temp.

room temp

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Meets ACS Specifications GR ACS

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay: 60.0-62.0%
Chloride (Cl): 0.001% max
Color (APHA): 10 max
Form: Liquid
Heavy metals (as Pb): 1ppm max
Iron (Fe): 1ppm max
Nitrogen compounds (as N): 0.001% max
Residue after ignition: 0.003% max
Silicate and phosphate (as SiO2): 5ppm max
Sulfate (SO4): 0.001% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service