Skip to Content
Merck

Skip To

PLBC09005

Regenerated Cellulose Membrane Filter, 3 kDa NMWCO

Ultracel®, filter diam. 90 mm

Synonym(s):

Ultrafiltration Discs, 3 kDa NMW, Regenerated Cellulose Membrane Filter, 3 kDa NMWCO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104903
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
3 kDa NMWCO
Material:
regenerated cellulose membrane
Feature:
hydrophilic
Diam.:
90 mm
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ultracel® 3 kDa Ultrafiltration Discs, Chemistry: Regenerated Cellulose (RC) Filter Diameter: 90 mm Amicon® Bioseparations

material

regenerated cellulose membrane

Quality Segment

feature

hydrophilic

manufacturer/tradename

Ultracel®

technique(s)

protein clean-up: suitable (desalting), protein purification: suitable (concentration)

diam.

90 mm

filter diam.

90 mm

filtration area

63 cm2

matrix

Ultracel®

pore size

3 kDa NMWCO

shipped in

ambient

General description

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel regenerated cellulose and Biomax polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore PVDF cut discs are available 0.1 µm, 0.22 µm, and 0.45 µm pore sizes in both 63.5 mm and 75 mm diameters.

Features & Benefits:
•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
•Both Ultracel, Durapore, and Biomax membranes are available in a wide range of pore sizes to meet your separation requirements
•Compatible with Amicon Stirred Cells

Applications:
Concentration, Buffer Exchange, Desalting
Device Configuration: Ultrafiltration Discs
Filter Code: PLBC
Filter Type: Ultrafiltration

Application

3 kDa NMWL, Ultracel regenerated cellulose, 90 mm diameter

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PLBC15005PLBC02510PLBC04310
material

regenerated cellulose membrane

material

regenerated cellulose membrane

material

regenerated cellulose membrane

material

regenerated cellulose membrane

diam.

90 mm

diam.

150 mm

diam.

25 mm

diam.

44.5 mm

pore size

3 kDa NMWCO

pore size

3 kDa NMWCO

pore size

3 kDa NMWCO

pore size

3 kDa NMWCO

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

matrix

Ultracel®

matrix

Ultracel®

matrix

Ultracel®

matrix

Ultracel®


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters