Skip to Content
Merck

PCC030C10G

Pellicon® Capsule with Ultracel® Membrane

pore size 30 kDa, C Screen, 1 m2 manifold with AseptiQuik® G connectors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903

material

EPDM rubber seal
PPO/PS blend housing
TPE seal
cellulose screen (Composite Regenerated)
composite regenerated cellulose (CRC) membrane
epoxy adhesive
epoxy device (Potting, BPA-free)
polyester screen
polypropylene screen
polyurethane adhesive
polyurethane device (Pottin)

Quality Level

description

C Screen, 1 m2 manifold with AseptiQuik® G connectors

sterility

sterile; γ-irradiated

feature

holdup volume 254 mL (in feed channel)
holdup volume 306 mL (in permeate channel)

parameter

max. inlet pressure at 75 psi (5.1 bar) and 4-30 °C
max. transmembrane pressure at 50 psi (3.5 bar) and 4-30 °C
4-8 L/min-m2 flow rate

technique(s)

ultrafiltration: suitable

capsule L × W × H

19.56 cm (7.7 in.) × 34.37 cm (13.53 in.) × 40.46 cm (15.93 in.)

membrane area

1 m2

screen H

15.9 in. (40.5 cm)

screen L

7.7 in. (19.6 cm) , Length

screen W

13.5 in. (34.4 cm)

impurities

≤5 ppm Meets TOC (in retentate effluent, after a WFI flush of 20 L/m2)

matrix

Ultracel®

pore size

30 kDa

pH range

2-13

fitting

inlet/outlet connection (Feed: AseptiQuik® G Connector)
inlet/outlet connection (Permeate: AseptiQuik® G Connector)
inlet/outlet connection (Retentate: AseptiQuik® G Connector)

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The Pellicon® Capsule with Ultracel Membrane is the first of its kind -- a true single-use TFF device that comes ready to process in minutes. Self-contained, it employs an easy-to-use, holderless format and is supplied gamma sterilized with preservative-free reverse osmosis water. This new addition to the Pellicon® family shares the same unbeatable performance of Pellicon® cassettes featuring C Screen, 1 m² manifold with AseptiQuik® G connectors. This device is linearly scalable, making it easy to transition between both formats.

Pellicon® Capsules are ideal for the filtration of solutions that require single-use capabilities, including:

-Rapid batch turnaround for higher plant productivity
-Lower risks of product cross-contamination
-Self-contained device for increased operator safety
-Process flexibility for multi-product manufacturing facilities
-Optimum use of time and resources from minimal device preparation

Membrane

Pellicon® Capsules with Ultracel membrane are designed for excellent product retention, recovery, and high yields. Ultracel void-free composite membranes combine ultra-low protein binding, low fouling, and organic solvent resistance with superb mechanical strength.

Application

Concentration, Diafiltration, Buffer exchange, Protein purification

Features and Benefits

  • Plug ′n play, holderless design - easy to install, safe to remove
  • Gamma Sterilized and preseravtive-free - ready to process in minutes
  • True single-use, self-contained capusle - fast, safe, and flexible batch turnaround
  • Proven Ultracel® membrane and C screen - high recovery, superior mass transfer, solvent resistance
  • Pellicon® TFF proven performance - true linear scalability within Pellicon® TFF families

Packaging

A foam pouch is inserted over the end caps on both ends of the capsule which is then double-bagged and vacuum sealed. The double-bagged capsule is then placed on a tray and individually boxed.

Compatibility

Screen Type: Type C screen (coarse screen)

Other Notes

The Emprove® Program complements the product portfolio through

  • Comprehensive documentation to support qualification, risk assessment and process optimization needs.
  • Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
  • Convenient 24/7 access to up-to-date product information.

Legal Information

AseptiQuik is a registered trademark of Colder Products Company
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.

Related Content

Learn more on mAb downstream processing, more specifically concentration and diafiltration, and the relevant associated products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service