Skip to Content
Merck
All Photos(1)

Key Documents

NX0407

Supelco

Nitric Acid

OmniTrace

Synonym(s):

Nitric acid solution, Hydrogen Nitrate, Azotic Acid,Aqua Fortis, Aquafortis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352301
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

vapor pressure

9.4 hPa ( 20 °C)

Assay

69%

form

liquid

concentration

≥65-<70% in nitric acid

pH

<1 (20 °C in H2O)

bp

121 °C/1013 hPa

mp

-32 °C

solubility

soluble (Water: Soluble)

density

1.39 g/cm3 at 20 °C (68 °F)

shipped in

ambient

storage temp.

room temp

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Aqueous OmniTrace
NX0407-1 and NX0407-4. Product is packed into HDPE bottles. Each HDPE bottle is packed into a single bottle shipping case. Reassay date for HDPE bottle stock is 2 years, refer to the batch specific COA.
NX0407-2. Product is packed into a poly coated glass bottle. NX0407-2 will be discontinued when stock is exhausted.

Legal Information

OMNITRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

  1. Hi, May I know the metal content in NX0407-4 Nitric Acid 67 - 70% OmniTrace® before i placed an order?

    1 answer
    1. The exact amount of each trace metal in Product NX0407 is listed in the lot-specific Certificate of Analysis. The specifications for maximum amounts of each metal (and other impurities) can be found on the product detail page below:
      https://www.emdmillipore.com/product/Nitric-Acid,EMD_CHEM-NX0407

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service