This product is a normal mouse serum and may be used as an Fc blocker in flow cytometry applications. For more information, please see: https://www.sigmaaldrich.com/product/Normal-Mouse-Serum,EMD_BIO-NS03L
NS03L
Normal Mouse Serum
This Normal Mouse Serum is validated for use in ELISA, Flow Cytometry, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation for the detection of Mouse Serum.
Synonym(s):
Mouse Serum
About This Item
Recommended Products
biological source
mouse
form
lyophilized
contains
≤0.1% sodium azide as preservative
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
shipped in
ambient
storage temp.
2-8°C
General description
Application
Flow Cytometry (see comments)
Immunoblotting (see comments)
Immunofluorescence (see comments)
Immunohistochemistry (see comments)
Immunoprecipitation (see comments)
Warning
Physical form
Reconstitution
Other Notes
Legal Information
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is this product can be used as a replacement of Fc blocker in flow cytometry?
1 answer-
Helpful?
-
-
Which strain of mouse is used to derive the serum for product NS03L-1ML?
1 answer-
The serum with catalogue number NS03L-1ML, is pooled from one strain only: Outbred non-swiss white mice, which are also known as Albino mice.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service