Skip to Content
Merck

NM0042

NovaSeal Battery

Battery, 10.8 V Li-ion

Synonym(s):

Spare battery for NovaSeal battery powered crimping tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41121815

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

NovaSeal Battery, Battery, 10.8 V Li-ion

sterility

non-sterile

product line

NovaSeal

compatibility

for use with NovaSeal battery powered crimping tool

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CMP0415XX6320022NM0047
sterility

non-sterile

sterility

-

sterility

-

sterility

non-sterile

product line

NovaSeal

product line

-

product line

-

product line

NovaSeal

compatibility

for use with NovaSeal battery powered crimping tool

compatibility

-

compatibility

for use with Millipore Single/Dual Chamber Incubator

compatibility

for use with NovaSeal battery powered crimping tool - NM0044

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service