ND-2ML-B
Anti-P1 (clone P3NIL100), Blood Grouping Reagent
BIOSCOT™ Anti-P1 from the clone P3NIL100, Human IgM, Blood Grouping Reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
human
Quality Level
clone
P3NIL100, monoclonal
packaging
pkg of 2 mL
manufacturer/tradename
BIOSCOT™
storage condition
do not freeze
concentration
0.5-3 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6-8
isotype
IgM
application(s)
clinical testing
shipped in
ambient
storage temp.
2-8°C
General description
Contains 0.1% (w/v) Sodium Azide
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months
Product formulation conforms to the requirements of the EU IVD directive.
For In Vitro use only
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months
Product formulation conforms to the requirements of the EU IVD directive.
For In Vitro use only
Application
For use by Tube technique
Vialled, Labeled
Legal Information
BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service