Skip to Content
Merck

Skip To

MXPPLUVHP

Milliflex® PLUS Pump

The Milliflex® PLUS vacuum pump is suitable for use in isolators. The pump is Vapor Hydrogen Peroxide (VHP) resistant and compatible, suitable for sterility testing

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NB.79
UNSPSC Code:
23151818
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

input

sample type pharmaceutical(s)

application(s)

bioburden testing
pharmaceutical
water monitoring, sterility testing

compatibility

for use with Milliflex® Systems, standard for VHP

shipped in

ambient

General description

The compact Milliflex® PLUS VHP vacuum pump streamlines filtration steps during testing to accelerate and ease the sample preparation. When used with convenient Milliflex® filtration funnels, the pump and Milliflex® units comprise an integrated solution for delivering reliable and accurate results. The pump′s small footprint suits tight workspaces and its low profile allow easy access and usage in isolators. The housing and the external parts are VHP resistant.

Application

The VHP Resistant Milliflex® PLUS Pump is designed to perform filtrations of samples inside an isolator. It is suitable for rapid sterility testing in combination with the Milliflex® Rapid System.

Features and Benefits

  • Streamlines filtration steps
  • Can be used with the Milliflex® filtration funnels
  • Compatible with the isolator environment and VHP decontamination
  • Compatible with the Milliflex® Rapid System to perform rapid sterility testing in a clean environment (isolator)

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Questions

Reviews

No rating value

Active Filters