Skip to Content
Merck

Skip To

MXPPLUS03

Milliflex Plus Pump Triple Head Kit

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, Water Monitoring

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NB.79
UNSPSC Code:
41116129
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Milliflex®

input

sample type bottled water
sample type pharmaceutical(s)
sample type beer
sample type cosmetics

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

shipped in

ambient

General description

The compact Milliflex Plus vacuum pump streamlines filtration to accelerate QC testing. Together, the Milliflex filter units and the Milliflex Plus vacuum pump provide an integrated solution to deliver reliable and accurate results in high throughput production environments.
Sanitization is easy, so there’s minimal disruption to workflow. For even greater throughput, a high-volume workstation of up to three Milliflex PLUS pumps can be set-up and used by a single operator. Testing reveals 3 Milliflex PLUS pumps are faster than a 6-place manifold.

Support and Services
{HCompany} provides technical support and a comprehensive services offering to help you save your QC resources and facilitate the implementation of your MilliFlex Plus Pump Triple unit into your daily testing routine. Our highly experienced teams are there to assist you.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content





Questions

Reviews

No rating value

Active Filters