Skip to Content
Merck

Skip To

MXPESP5EP

EPDM O-ring + Plug Seal

Accessory for On-line sampling and filtration of aqueous liquids

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24
eCl@ss:
32160606
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

EPDM rubber

Quality Segment

packaging

pkg of 5 numbers

manufacturer/tradename

MicropreSURE®

input

sample type beer
sample type wine
sample type food(s)
sample type sport drink
sample type dairy
sample type soft drink
sample type bottled water
sample type pharmaceutical(s)
sample type beverage(s)
sample type cider

application(s)

bioburden testing
food and beverages
pharmaceutical

shipped in

ambient

General description

EPDM O-ring + plug seal is an accessory of the MicropreSure® sampler. MicropreSure® sampler is an on-line filtration sampler to simplify in-process microbiological testing. It is a simple, protective design that allows samples to be aseptically processed right at the collection site thereby, reducing contamination risk. The EPDM rubber O-ring + plug seal aids in the sealed and leak-free flow of the sample.

Application

EPDM O-ring + plug seal is used for in-process sampling and filtration of aqueous liquids such as beer, dairy, bottled water, pharmaceutical products analysis, soft drink, cider, sports drink, and wine for bioburden testing.

Legal Information

MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MXPESP5VTMXPESP50REZFITGASK3
material

EPDM rubber

material

fluoroelastomer

material

silicone

material

EPDM rubber

packaging

pkg of 5 numbers

packaging

pkg of 5 numbers

packaging

pkg of 5 numbers

packaging

pkg of 3 pieces

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

application(s)

bioburden testing
food and beverages
pharmaceutical

application(s)

bioburden testing
food and beverages
pharmaceutical

application(s)

bioburden testing
food and beverages
pharmaceutical

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

input

sample type beer
sample type wine
sample type food(s)
sample type sport drink
sample type dairy
sample type soft drink
sample type bottled water
sample type pharmaceutical(s)
sample type beverage(s)
sample type cider

input

sample type wine
sample type dairy
sample type cider
sample type pharmaceutical(s)
sample type beer
sample type food(s)
sample type sport drink
sample type bottled water
sample type soft drink
sample type beverage(s)

input

sample type pharmaceutical(s)
sample type dairy
sample type beverage(s)
sample type cider
sample type wine
sample type sport drink
sample type beer
sample type bottled water
sample type soft drink
sample type food(s)

input

-

Quality Level

SP1

Quality Level

SP1

Quality Level

EQ1

Quality Level

SP1




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters