Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
1-Methyl-2-pyrrolidone, Biosynthesis OmniSolv®
vapor pressure
0.32 hPa ( 20 °C)
Quality Segment
assay
≥99.7% (GC)
form
liquid
dilution
(for synthesis)
impurities
≤1.0 μeq/g Titrable base, ≤200 ppm Water (H2O), ≤5 ppm Amines (as dimethylamine)
color
APHA: ≤10
pH
8.5-10.0 (20 °C, 100 g/L in H2O)
bp
202 °C/1013 hPa
mp
-24 °C
transition temp
flash point 91 °C
solubility
1000 g/L
density
1.03 g/cm3 at 20 °C
UV cutoff
≤270 nm
UV absorption
λ: 270 nm Amax: ≤1.00, λ: 280 nm Amax: ≤0.50, λ: 300 nm Amax: ≤0.10, λ: 325 nm Amax: ≤0.05, λ: 350 nm Amax: ≤0.03, λ: 400 nm Amax: ≤0.005
shipped in
ambient
storage temp.
room temp
SMILES string
N1(CCCC1=O)C
InChI
1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3
InChI key
SECXISVLQFMRJM-UHFFFAOYSA-N
Application
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Preparation Note
Analysis Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99.7% (GC) | assay ≥99.7% (GC) | assay ≥99.5% (GC) | assay ≥99.8% (GC) |
| Quality Level 200 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| solubility 1000 g/L | solubility 1000 g/L | solubility 1000 g/L | solubility 1000 g/L |
| form liquid | form liquid | form liquid | form liquid |
| pH 8.5-10.0 (20 °C, 100 g/L in H2O) | pH 8.5-10.0 (20 °C, 100 g/L in H2O) | pH 8.5-10.0 (20 °C, 100 g/L in H2O) | pH 8.5-10.0 (20 °C, 100 g/L in H2O) |
| UV absorption λ: 270 nm Amax: ≤1.00, λ: 300 nm Amax: ≤0.10, λ: 350 nm Amax: ≤0.03, λ: 280 nm Amax: ≤0.50, λ: 400 nm Amax: ≤0.005, λ: 325 nm Amax: ≤0.05 | UV absorption λ: 270 nm Amax: ≤1.00, λ: 300 nm Amax: ≤0.20, λ: 325 nm Amax: ≤0.10, λ: 350 nm Amax: ≤0.03, λ: 400 nm Amax: ≤0.005 | UV absorption - | UV absorption - |
Still not finding the right product?
Explore all of our products under 1-Methyl-2-pyrrolidone
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 1
flash_point_f
195.8 °F - Pensky-Martens closed cup
flash_point_c
91 °C - Pensky-Martens closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The 4L M-Bottle provides safe handling of solvents due to an innovative solvent bottle design with sealing technology to eliminate leaks and reduce risks in solvent dispensing.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MX1392-1 | 04061839430082 |
| MX1392P-1 | 04061839430150 |
| MX1392-31 | 04061839430099 |
| MX1392-5 | 04061839430143 |



