Skip to Content
Merck

Skip To

MX1301

4-Methyl-2-pentanone

OmniSolv®

4-Methyl-2-pentanone

Synonym(s):

MIK, Hexone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
EC Index Number:
203-550-1
UNSPSC Code:
12352115
Beilstein/REAXYS Number:
605399
MDL number:
Assay:
≥99% (GC)
Technique(s):
HPLC: suitable, LC/MS: suitable, gas chromatography (GC): suitable
Bp:
117-118 °C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.5 (vs air)

Quality Level

product line

OmniSolv®

assay

≥99% (GC)

form

liquid

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

technique(s)

HPLC: suitable, LC/MS: suitable, gas chromatography (GC): suitable

impurities

≤0.08% Water (H2O)

evapn. residue

≤5 ppm

color

APHA: ≤10

refractive index

n20/D 1.395 (lit.)

pH

7 (20 °C)

bp

117-118 °C

mp

−80 °C (lit.)

transition temp

flash point 14 °C

solubility

14.1 g/L

density

0.8 g/cm3 at 20 °C, 0.801 g/mL at 25 °C (lit.)

UV cutoff

≤334 nm

UV absorption

λ: 334 nm Amax: ≤1.00, λ: 339 nm Amax: ≤0.50, λ: 349 nm Amax: ≤0.20, λ: 400 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

General description

4-Methyl-2-pentanone also known as MIBK, is widely used in analytical chemistry as a solvent due to its excellent volatility, low water solubility, and compatibility with both polar and non-polar analytes. In chromatography, MIBK is particularly valued for its role in gas chromatography (GC) and sample preparation workflows, where it enhances analyte recovery and resolution in complex matrices.

Application

  • MIBK is used as a solvent and diluent in GC analysis of volatile organic compounds (VOCs), hydrocarbons, and environmental pollutants.
  • MIBK has well-characterized retention indices and vapor-phase behavior, making it suitable for use in GC method development and calibration
OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
is packaged in 4L amber glass bottle

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.0% minColor (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 334 nm1.00AU maxUV Abs. at 339 nm0.50AU maxUV Abs. at 349 nm0.20AU maxUV Abs. at 400 nm0.01AU maxUV Cut-off334nm maxWater (H2O)0.08% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
36051153771302474
Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

assay

≥99% (GC)

assay

≥98.5%

assay

≥99%

assay

≥99.7% (GC)

technique(s)

HPLC: suitable, gas chromatography (GC): suitable, LC/MS: suitable

technique(s)

-

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

solubility

14.1 g/L

solubility

-

solubility

water: soluble 20 g/L

solubility

-

form

liquid

form

liquid

form

-

form

-

bp

117-118 °C

bp

117-118 °C

bp

117-118 °C

bp

117-118 °C


Still not finding the right product?

Explore all of our products under 4-Methyl-2-pentanone


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

57.2 °F - closed cup

flash_point_c

14 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MX1301-104061839430044

Questions

Reviews

Active Filters

  1. Cottage Grove, MN
    • Review 1
    • Votes 0
    2 out of 5 stars.

    C of A's for EMD chemicals

    What happened to the C of A's that had actual Lot results in them? The C of A now is more like a product spec sheet, not a real C of A. I'm not pleased!

    Helpful?

    1. Response from Merck KGaA:

      Thank you for your review. We are sorry to hear that your experience did not match your product expectations. We encourage customers who experience problems with our products to contact us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.