Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
agency
meets USP/NF
Quality Segment
vapor density
3.5 (vs air)
assay
≥98.5% (GC)
form
liquid
autoignition temp.
840 °F
expl. lim.
1.2-8 %, 93 °F
dilution
(for analytical testing)
impurities
≤0.002 meq/g Titrable acid, ≤0.1% Water (H2O)
evapn. residue
≤0.005%
color
clearAPHA: ≤15
refractive index
n20/D 1.395 (lit.)
pH
7 (20 °C)
bp
117-118 °C
mp
−80 °C (lit.)
transition temp
flash point 14 °C
solubility
14.1 g/L
density
0.8 g/cm3 at 20 °C, 0.801 g/mL at 25 °C (lit.)
shipped in
ambient
storage temp.
room temp
SMILES string
CC(C)CC(C)=O
InChI
1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3
InChI key
NTIZESTWPVYFNL-UHFFFAOYSA-N
General description
Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.
Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.
Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.
Application
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Analysis Note
Assay (GC): 98.5% min
Color (APHA): 15 max
Evaporation residue: 0.005% max
Form: Liquid
Titrable acid: 0.002meq/g max
Water (H2O): 0.1% max
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 100 | Quality Level 300, 400 | Quality Level 100 |
| assay ≥98.5% (GC) | assay ≥99.5% | assay ≥99% | assay ≥99% |
| solubility 14.1 g/L | solubility - | solubility - | solubility water: soluble 20 g/L |
| form liquid | form liquid | form liquid | form - |
| expl. lim. 1.2-8 %, 93 °F | expl. lim. 1.2-8 %, 93 °F | expl. lim. 1.2-8 %, 93 °F | expl. lim. 1.2-8 %, 93 °F |
| refractive index n20/D 1.395 (lit.) | refractive index n20/D 1.395 (lit.) | refractive index n20/D 1.395 (lit.) | refractive index n20/D 1.395 (lit.) |
Still not finding the right product?
Explore all of our products under 4-Methyl-2-pentanone
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
57.2 °F - closed cup
flash_point_c
14 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MX1300-3 | 04061839430020 |
| MX1300-30 | 04061839430037 |



