Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
HPLC grade
Quality Segment
vapor pressure
268 hPa ( 20 °C)
product line
OmniSolv®
assay
≥99.5% (GC)
form
liquid
technique(s)
HPLC: suitable, LC/MS: suitable, gas chromatography (GC): suitable
impurities
≤0.05% Water (H2O), ≤1 ppm Peroxide (as H2O2), ≤5 ng/L Capillary ECD responsive substances (as PCNB)
evapn. residue
≤1 ppm
color
APHA: ≤10
bp
55.3 °C/1013 hPa
mp
-108.6 °C
transition temp
flash point -28 °C
solubility
42 g/L
density
~0.74 g/cm3 at 20 °C
UV absorption
λ: 210 nm Amax: ≤0.75, λ: 220 nm Amax: ≤0.50, λ: 225 nm Amax: ≤0.30, λ: 250 nm Amax: ≤0.10, λ: 270 nm Amax: ≤0.01, λ: 300 nm Amax: ≤0.005
application(s)
PFAS testing
shipped in
ambient
storage temp.
room temp
SMILES string
O(C(C)(C)C)C
InChI
1S/C5H12O/c1-5(2,3)6-4/h1-4H3
InChI key
BZLVMXJERCGZMT-UHFFFAOYSA-N
General description
Application
- A solvent in organic synthesis, particularly in reactions that require a non-polar medium, such as Grignard reactions and other nucleophilic substitutions.
- An effective extraction solvent for isolating organic compounds from natural products, allowing for the recovery of essential oils, flavors, and fragrances.
- A mobile phase in high-performance liquid chromatography (HPLC), especially in the separation of polar and non-polar compounds.
- A chemical intermediate in the production of various specialty chemicals and pharmaceuticals, contributing to the synthesis of complex molecules.
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Preparation Note
Analysis Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description OmniSolv® | description for liquid chromatography LiChrosolv® | description EMPLURA® | description for analysis EMSURE® ACS |
| grade HPLC grade | grade - | grade - | grade ACS reagent |
| application(s) PFAS testing | application(s) PFAS testing | application(s) - | application(s) - |
| technique(s) HPLC: suitable, gas chromatography (GC): suitable, LC/MS: suitable | technique(s) HPLC: suitable | technique(s) - | technique(s) - |
| assay ≥99.5% (GC) | assay ≥99.8% (GC) | assay ≥99% (GC) | assay ≥99.5% (GC) |
| Quality Level 100 | Quality Level 100 | Quality Level 200 | Quality Level 300 |
| solubility 42 g/L | solubility 42 g/L | solubility 42 g/L | solubility - |
Still not finding the right product?
Explore all of our products under tert-Butyl methyl ether
signalword
Danger
hcodes
Hazard Classifications
Flam. Liq. 2 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
flash_point_f
-18.4 °F - closed cup
flash_point_c
-28 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




