Skip to Content
Merck

Skip To

MX0760

2-Methylbutane

Practical

2-Methylbutane

Synonym(s):

2-Methylbutane, Isopentane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHCH2CH3
CAS Number:
UNSPSC Code:
12191502
EC Index Number:
201-142-8
MDL number:
Assay:
≥95% (GC)
Bp:
28 °C/1013 hPa
Vapor pressure:
800 hPa ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

800 hPa ( 20 °C)

Quality Level

assay

≥95% (GC)

form

liquid

color

APHA: ≤30

bp

28 °C/1013 hPa

mp

-160 °C

transition temp

flash point -57 °C

solubility

0.048 g/L

density

0.62 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

C(CC)(C)C

InChI

1S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3

InChI key

QWTDNUCVQCZILF-UHFFFAOYSA-N

General description

EMPARTA solvents are designed for routine chemical synthesis, drying, purification, and critical labware cleaning in quality control environments. Engineered for precision, they deliver consistently reproducible results essential for rigorous validation and documentation.

Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.

Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.

Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.

Application

2-Methylbutane can be used as a reactant to synthesize:
  • Isoprene by two-stage dehydrogenation process using chromia-alumina catalyst.
  • Alkyl aromatic compounds via silver-catalyzed selective C(sp3)–H benzylation reaction in the presence of N-triftosylhydrazones as carbene precursor.

It can also be used as a weakly polar solvent to study the intramolecular charge transfer (ICT) effect and the solvatochromic behavior of the pyrene substituted 1,3,4-oxadiazole derivatives.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Assay (GC): 95% min
Color (APHA): 30 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
27034259070M32631
Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100

assay

≥95% (GC)

assay

≥99.5%

assay

≥99.5% (GC)

assay

≥99%

solubility

0.048 g/L

solubility

-

solubility

-

solubility

-

form

liquid

form

liquid

form

liquid

form

liquid

density

0.62 g/cm3 at 20 °C

density

0.62 g/mL at 25 °C (lit.)

density

0.62 g/mL at 25 °C (lit.)

density

0.62 g/mL at 25 °C (lit.)

bp

28 °C/1013 hPa

bp

30 °C (lit.)

bp

30 °C (lit.)

bp

30 °C (lit.)


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

-59.8 °F - closed cup

flash_point_c

-51 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Site-selective C-H benzylation of alkanes with N-triftosylhydrazones leading to alkyl aromatics
Liu Zhaohong, et al.
Chem, 6(8) (2020)
Power-efficient synthesis of isoprene via two-stage dehydrogenation of isopentane
Karimov OK, et al.
World Applied Sciences Journal , 24(3) (2013)
Synthesis, characterization and photophysical properties of a new class of pyrene substituted 1, 3, 4-oxadiazole derivatives
Najare Mahesh S, et al.
Optical Materials, 88 (2019)



Global Trade Item Number

SKUGTIN
MX0760-104061839429819

Questions

Reviews

Active Filters

    • Review 1
    • Votes 0
    1 out of 5 stars.

    Helpful?

    1. Response from Merck KGaA:

      Thank you for your rating. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.