Skip to Content
Merck

Skip To

MX0160

Maltose, Monohydrate, Powder

Synonym(s):

Maltose, Monohydrate, Powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]([C@H](O)[C@@H](O)C=O)[C@H](O)CO.O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

General description



Specifications:
Loss on drying: 7% max
Melting range: 109-113°C
Specific rotation [a]25/D: +111.7-+130.4°





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.059111.059121.05910
Quality Level

100

Quality Level

500

Quality Level

100

Quality Level

200

form

powder

form

solid

form

solid

form

-

color

white

color

-

color

-

color

-

storage condition

OK to freeze

storage condition

-

storage condition

-

storage condition

-

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-


Still not finding the right product?

Explore all of our products under Maltose, Monohydrate, Powder


flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

11 - Combustible Solids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MX0160-2004055977229479
MX0160-104055977230314
MX0160-704055977229486

Questions

Reviews

No rating value

Active Filters