Skip to Content
Merck

Skip To

MX0100

Maleic acid

Synonym(s):

Butenedioic Acid, cis-Butenedioic acid, Toxilic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
NACRES:
NA.25
UNSPSC Code:
12352106
Beilstein/REAXYS Number:
605762
MDL number:
Assay:
≥98%
Form:
crystalline powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98%

Quality Segment

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

130-135 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

General description



Assay: ≥98%
Fumaric acid: ≤2%
Infrared spectrum: Conforms to standard





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

<ul>
<li><strong>Hydrophilization and Functionalization of Fullerene C(60) with Maleic Acid Copolymers by Forming a Non-Covalent Complex.</strong>: This study investigates the interaction between fullerene C(60) and maleic acid copolymers, focusing on enhancing the solubility and functionality of fullerene for potential biochemical applications (Samoilova et al., 2024).</li>
<li><strong>Nanodisc Technology: Direction toward Physicochemical Characterization of Chemosensory Membrane Proteins in Food Flavor Research.</strong>: This review discusses the role of nanodisc technology in food science, where maleic acid copolymers are utilized to stabilize membrane proteins for analytical studies (Karanth et al., 2024).</li>
</ul>

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL801428.17058M0100000
assay

≥98%

assay

≥98.0% (HPLC)

assay

99.0-101.0% substance (anhydrous) basis

assay

-

Quality Level

100

Quality Level

-

Quality Level

500

Quality Level

-

form

crystalline powder

form

powder

form

solid

form

-

storage temp.

room temp

storage temp.

-

storage temp.

15-25°C

storage temp.

-

mp

130-135 °C (lit.)

mp

130-135 °C (lit.)

mp

130-135 °C (lit.)

mp

130-135 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Maleic acid


pictograms

CorrosionExclamation mark

signalword

Danger

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MX0100-2004055977230307

Questions

Reviews

No rating value

Active Filters