Skip to Content
Merck
All Photos(1)

Key Documents

MSPNNFX00

Millipore

Multiscreen® 96 well Plate, solid bottom

flat bottom natural polypropylene plate, non-sterile, pkg of 100 plates

Synonym(s):

MultiScreen® 96-well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.15
material:
flat bottom natural polypropylene plate
size:
96 well
sterility:
non-sterile
Pricing and availability is not currently available.

material

flat bottom natural polypropylene plate

Quality Level

sterility

non-sterile

product line

MultiScreen®

packaging

pkg of 100 plates

plate size

96 wells

size

96 well

Looking for similar products? Visit Product Comparison Guide

General description

MultiScreen® 96-well polypropylene microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices. Polypropylene microplates are appropriate for sample preparation and storage in addition to some biochemical assays.

Physical form

Color Code:Natural

Other Notes

Automation Compatible:Yes

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. MSPNNFX00 Millipore Multiscreen® 96 well Plate, solid bottom Are these plates suitable for fluorescence assay? I am looking for plates compatible with 1 M sulfuric acid for fluorescence assay. Please suggest which ones are the best. Thank you!

    1 answer
    1. Polypropylene is not fully resistant to strong acids like 1 M sulfuric acid, so these plates are not recommended for use. Materials such as PTFE or HDPE are needed for full chemical resistance, and there are no plates made of these materials available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service