MPM0PS
MOPS SDS running buffer powder for mPAGE® Bis-Tris gels
pack of 5 ea (Each packet makes 1 liter of running buffer)
Synonym(s):
SDS running buffer
About This Item
Recommended Products
form
powder
packaging
pack of 5 ea (Each packet makes 1 liter of running buffer)
General description
- Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Application
Reconstitute 1 pouch of buffer powder with 1000 ml deionized water to make 1X running buffer. Do not use acid or base to adjust the pH.
- Western Blotting
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Explore the impact of water quality on protein electrophoresis and Western blotting. Learn about the role of ultrapure water and its effect on chemiluminescent signals in Western blotting.
Bis-Tris SDS-PAGE gel casting kits with mPAGE® TurboMix technology for rapid, reliable gel casting and protein separation.
Protocols
Instructions for setting up an mPAGE™ SDS-PAGE precast protein gel for gel electrophoresis using an Invitrogen XCell Surelock® mini-cell.
Instructions for setting up an mPAGE SDS-PAGE precast protein gel for gel electrophoresis using a Bio-Rad Mini-PROTEAN® electrophoresis system.
Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.
Related Content
mPAGE® gels offer high-resolution protein separation, reduced cost, and compatibility with common electrophoresis tanks.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service