Skip to Content
Merck
All Photos(1)

Key Documents

MMQTV0KT1

Milliflex Oasis® Quantum Kit

Including Milliflex Oasis® HAWG funnels, parameter 100 mL, pore size 0.45 μm, (White gridded membrane)

Synonym(s):

Fluorescence-based rapid detection system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

material

mixed cellulose esters (MCE)

manufacturer/tradename

Milliflex® Quantum

parameter

100 mL

volume

47 mm

pore size

0.45 μm

General description

Milliflex® Quantum Universal Kit for Milliflex Oasis® is a fluorescence-based technology for the quantitative and rapid detection of microorganisms in filterable samples. The universal enzymatic fluorescent staining is non-destructive, allowing the identification of the contaminant following a positive result. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms. The kit includes fluorescent reagents and Milliflex Oasis® HAWG 100 mL funnels (0.45 μm membrane, White gridded).

Application

The Milliflex® Quantum Universal Kit for Milliflex Oasis® is used for rapid bioburden testing in filtrable samples, including in-process and final product samples (pharmaceuticals and water). The product is also used in industrial QC and water monitoring.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service