MANTA4TR1
MAS-100 NT® Test Record Protocol
configured for MAS-100 NT® (A4 format)
About This Item
Recommended Products
material
paper (A4)
compatibility
configured for MAS-100 NT® (A4 format)
General description
With our best-in-class services you can:
- Optimize your QC lab workflow and ensure regulatory compliance.
- Rely on comprehensive and ready-to-use validation packages.
- Ensure the performance of your MAS-100 NT portable air sampler while reducing the risk of breakdown.
Application
Features and Benefits
- Validation Master Plan: Define structure, responsibilities for qualification.
- Installation Qualification (IQ)
Verification of product′s utilities and operating environment requirements.
Equipment and personnel preparation.
- Operational Qualification (OQ): Verification of product′s functionality (hardware, software, devices).
- Performance Qualification (PQ) Test Method suitability verification (microbiology validation procedures).
- Final Report Summarizes all testing performed for final approval of validation.
Other Notes
Legal Information
accessory