Skip to Content
Merck
All Photos(1)

Key Documents

MAC138

Sigma-Aldrich

Anti-Inntag 5 (IT5) Antibody, clone R19/8-11/18

clone R19/8-11/18, from mouse

Synonym(s):

Pollen allergen Phl p 2, Phl p2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

R19/8-11/18, monoclonal

species reactivity

Phleum pratense

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Pollen allergen Phl p 2 (UniProt: P43214; also known as Allergen Phl p II, Phl p 2) is encoded by the PHLPII gene in Phleum pratense (Common timothy). Phl p2 binds to IgE and causes allergic reactions in human. Inntag 5 IT5 is a small protein domain of Phelum pratense that display strong structural determinants and does not generate solubility issues in vivo and in vitro. Also, it is not shown to compromise cell fitness when overexpressed and has no detectable functional and localization effects when fused to a variety of target proteins. Although the Inntag IT5 has a similar length to that of the Flag tag, it does not affect cell morphology and produces only very mild effect in the mean critical size at the G1/S transition compared to Flag tag. The structure of Inntag IT5 is based on a packed beta-sheet confirmation.

Specificity

Clone R19/8-11/18 detects Pollen allergen Phl p 2 in lysate from HEK293 cells transfected with GFP-IT5.

Immunogen

His-tagged recombinant fragment corresponding to 96 amino acids of Pollen allergen Phl p 2 chain.

Application

Anti-Inntag 5 (IT5), clone R19/8-11/18 is a mouse monoclonal antibody that detects the Innocuous Inntag IT5 based on pollen allergen PhIp2 sequence and is tested for Immunofluorescence, Immunoprecipitation, and Western Blotting.
Research Category
Secondary & Control Antibodies
Western Blotting Analysis: A representative lot detected Inntag 5 (IT5) in Western Blotting applications (Georgieva, M.V., et. al. (2015). Nat Methods. 12(10):955-8).

Immunoprecipitation Analysis: A representative lot detected Inntag 5 (IT5) in Immunoprecipitation applications (Georgieva, M.V., et. al. (2015). Nat Methods. 12(10):955-8).

Immunofluorescence Analysis: A representative lot detected Inntag 5 (IT5) in Immunofluorescence applications (Georgieva, M.V., et. al. (2015). Nat Methods. 12(10):955-8).

Quality

Evaluated by Western Blotting in transfected HEK293/GFP-IT5 cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Inntag 5 (IT5) in 10 µg of transfected HEK293/GFP-IT5 cell lysates.

Target description

~40.2 kDa observed; 13.36 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service