Skip to Content
Merck

Skip To

MAC115

Anti-human Albumin Antibody, clone 8F6F9

ascites fluid, clone 8F6F9, from mouse

Synonym(s):

Serum albumin, ALB, human albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
8F6F9, monoclonal
Application:
WB
Citations:
1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

8F6F9, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ALB(213)

General description

Human Albumin, also known as Serum albumin (ALB), is a member of the ALB/AFP/VDB family. Serum albumin is the main protein of plasma and has been shown to bind with water, Ca2+, Na+, K+, fatty acids, hormones, bilirubin and drugs. Human Albumin plays an important role in the regulation of the colloidal osmotic pressure of the blood. Additionally, Human Albumin has been shown to be the major zinc transporter in plasma, and typically binds about 80% of all plasma zinc. Mutations in ALB are implied to be the cause of familial dysalbuminemic hyperthyroxinemia (FDH), a form of euthyroid hyperthyroxinemia that is due to increased affinity of ALB for T4. Human Albumin is expressed primarily in plasma.
~50 and ~69 observed. Uncharacterized bands may appear in some lysate(s).

Immunogen

Human sera albumin

Application

Detect Albumin using this mouse monoclonal antibody, Anti-human Albumin Antibody, clone 8F6F9 validated for use in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Human serum and plasma
Evaluated by Western Blotting in human serum and plasma.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected human Albumin in human serum and plasma.

Optimal working dilutions must be determined by end user.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6075SRP6516
biological source

mouse

biological source

mouse

biological source

human

clone

8F6F9, monoclonal

clone

OVA-14, monoclonal

clone

-

Gene Information

human ... ALB(213)

Gene Information

chicken ... Alb(396197)

Gene Information

human ... ALB(213)

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

-

species reactivity

human

species reactivity

chicken, turkey

species reactivity

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Naiyou Liu et al.
Biomedicines, 10(9) (2022-09-24)
Myofibroblast activation is a cellular response elicited by a variety of physiological or pathological insults whereby cells initiate a coordinated response intended to eradicate the insult and then revert back to a basal state. However, an underlying theme in various



Global Trade Item Number

SKUGTIN
MAC11504053252918582

Questions

Reviews

No rating value

Active Filters