Skip to Content
Merck
All Photos(1)

Key Documents

MABX5522

Sigma-Aldrich

Anti-CEA (10mg) KC

from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

Gene Information

human ... CEACAM5(1048)

General description

Carcinoembryonic antigen (CEA) is a protein associated with cell adhension in a human fetus. The production of CEA usually stops before birth therefore is only present at very low level in healthy adults. Elevated levels of CEA in serum can be associated with some cancers so the it is regularly used as a tumour marker in clinical tests.

Specificity

Specific for Human Carcinoembryonic Antigen. Negative for human albumin, NCA, NCA-2, and PSA, etc

Immunogen

corresponding to Human Carcinoembryonic antigen.

Application

Detect Carcinoembryonic antigen using this mouse monoclonal Anti-CEA (10mg) KC, Cat. No. MABX5522-10KC, validated for use in ELISA and Lateral Flow Assay.
Research Category
Apoptosis & Cancer
Suggested antibody pairs MABX5521-1KC and MABX5522-1KC

Quality

Tested

Physical form

Format: Purified
Ion-exchange chromatography
Purified mouse monoclonal in buffer containing 50 mM NaCl, 10 mM PBS, pH 8.0, 0.1 % sodium azide (NaN₃).

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What does KC means here?

    1 answer
    1. The 'KC' designation for this product, as well as a number of other antibodies, is intended to signify a bulk or custom quantity. Most antibodies are available in microgram quantities. Note that this item, as well as others, are available in a 1mg and a 10mg package size.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service