Skip to Content
Merck
All Photos(1)

Key Documents

MABX5512

Sigma-Aldrich

Anti-AFP (10mg) KC

from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

Gene Information

human ... AFP(174)

General description

Alpha-fetoprotein (AFP) is a protein present in a developing fetus. The production of AFP usually stops before birth therefore is only present at very low level in healthy non-pregnant adults. Elevated levels of AFP in serum can be an indication of certain cancers including liver, testicular and overian tumours so it is regularly used as a cancer marker in clinical tests.

Specificity

Specific for alpha -Fetoprotein

Immunogen

corresponding to Human Alpha-fetoprotein.

Application

Detect Alpha-fetoprotein using this mouse monoclonal Anti-AFP (10mg) KC, Cat. No. MABX5512-10KC, validated for use in ELISA and Lateral Flow Assay.
Research Category
Apoptosis & Cancer
Suggested antibody pairs MABX5511-1KC and MABX5512-1KC

Quality

Tested

Physical form

Format: Purified
Ion-exchange chromatography
Purified mouse monoclonal in buffer containing 50 mM NaCl, 10 mM PBS, pH 8.0, 0.1 % sodium azide (NaN₃).

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service