Skip to Content
Merck
All Photos(1)

Key Documents

MABT823

Sigma-Aldrich

Anti-alpha-Actin Antibody, Cardiac Muscle (ACTC1) Antibody

mouse monoclonal, 22D3

Synonym(s):

Actin, alpha cardiac muscle 1, Alpha-cardiac actin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-alpha-Actin Antibody, Cardiac Muscle Antibody, clone 22D3, clone 22D3, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

22D3, monoclonal

species reactivity

rat, human, goat, rabbit

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ACTC1(70)

General description

Actin, alpha cardiac muscle 1 (UniProt P68032; also known as Alpha-cardiac actin) is encoded by the ACTC1 (also known as ACTC, ASD5, CMD1R, CMH11, LVNC4) gene (Gene ID 70) in human. Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin is encoded by the ACTA1 gene, smooth muscle alpha-actin by the ACTA2 gene, cytoplasmic beta-actin by the ACTB gene, cardiac muscle alpha-actin by the ACTC1 gene, cytoplasmic gamma-actin by the ACTG1 gene, and smooth muscle gamma-actin by the ACTG2 (a.k.a. ACTA3) gene. Although actins show >90% overall sequence homology, isoforms do show spatial, temporal, and tissue-specific expresson patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic - and -actins, are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt.

Specificity

Clone 22D3 specifically detected alpha-cardiac actin in rat myocardium homogenates with no cross-reactivity toward - or -smooth muscle actin in rat aorta and chicken gizzard homogenates, cytoplasmic - or -actin in human platelet lysate, nor -skeletal actin in rat skeletal muscle homogenate (Driesen, R.B., et al. (2009). J. Cell. Mol. Med. 13(5):896-908).

Immunogen

KLH-conjugated linear peptide corresponding to human mature alpha-cardiac actin N-terminal end sequence with an acetylated N-terminus.

Application

Immunocytochemistry Analysis: A representative lot immunostained 4% paraformaldehyde-fixed, 0.05% NP-40-permeabilzed rectangular rabbit cardiomyocytes by indirect immuoflourescence (Driesen, R.B., et al. (2009). J. Cell. Mol. Med. 13(5):896-908).

Immunofluorescence Analysis: A representative lot immunostained cardiomyocytes in human left ventricular endocardial biopsies, rabbit left ventricular myocardium, as well as in goat atrial myocardium frozen tissue sections by fluorescent immunohistochemistry (Driesen, R.B., et al. (2009). J. Cell. Mol. Med. 13(5):896-908).

Western Blotting Analysis: A representative lot specifically detected alpha-cardiac actin in rat myocardium homogenate with no cross-reactivity toward - or -smooth muscle actin in rat aorta and chicken gizzard homogenates, cytoplasmic - or -actin in human platelet lysate, nor -skeletal actin in rat skeletal muscle homogenate (Driesen, R.B., et al. (2009). J. Cell. Mol. Med. 13(5):896-908).
Research Category
Cell Structure
This mouse monoclonal Anti-alpha-Actin Antibody, Cardiac Muscle, clone 22D3, Cat. No. MABT823 is validated for use in Flow Cytometry, Immunocytochemistry, Immunofluorescence, and Western Blotting for the detection of alpha-cardiac actin.

Quality

Evaluated by Flow Cytometry in HeLa cells.

Flow Cytometry Analysis: 2 µL of this antibody detected alpha-cardiac actin in one million 4% paraformaldehyde-fixed, 0.2% Triton X-100-permeabilzed HeLa cells.

Target description

41.78/42.02 kDa (mature/pro-form) calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rowan M Karvas et al.
Molecular human reproduction, 26(6), 425-440 (2020-05-03)
Human placental development during early pregnancy is poorly understood. Many conceptuses are lost at this stage. It is thought that preeclampsia, intrauterine growth restriction and other placental syndromes that manifest later in pregnancy may originate early in placentation. Thus, there
Christine Chaponnier et al.
F1000Research, 5, 416-416 (2016-06-24)
Higher vertebrates (mammals and birds) express six different highly conserved actin isoforms that can be classified in three subgroups: 1) sarcomeric actins, α-skeletal (α-SKA) and α-cardiac (α-CAA), 2) smooth muscle actins (SMAs), α-SMA and γ-SMA, and 3) cytoplasmic actins (CYAs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service