Skip to Content
Merck
All Photos(2)

Key Documents

MABT1366

Sigma-Aldrich

Anti-CRB3A Antibody, clone 1E6

clone 1E6, from rat

Synonym(s):

Protein crumbs homolog 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
1E6, monoclonal
application:
IHC
WB
species reactivity:
human
technique(s):
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:
1

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E6, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), Drosophila (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CRB3(92359)

Related Categories

General description

Protein crumbs homolog 3 (UniProt: Q9BUF7) is encoded by the CRB3 (also known as UNQ588/PRO1158) gene (Gene ID: 92359) in human. Mammalian cells contain three Crb orthologs. CRB1 is primarily expressed in the brain, cornea, and retina. CRB2 is expressed in lung, kidney, retina, colon and mammary glands. It is also expressed in at moderate levels in Expressed at moderate levels in liver, spleen, pancreas, placenta, and prostate tissue. CRB3 is present in epithelial tissues and is present in two isoforms, CRB3A and CRB3B. CRB3 is a single-pass type I membrane protein that is reported to control epithelial cell polarization. CRB3A is apically localized and promotes apical membrane growth and is and is essential for the formation of tight junctions in epithelial cells. It also prevents the epithelial-to-mesenchymal transition (EMT), which is known to contribute to tumor progression. CRB3 is synthesized with a signal peptide (aa 1-26), which is subsequently cleaved off. The mature form contains an extracellular domain (aa 27-59), a transmembrane domain (aa 60-80), and a cytoplasmic tail (aa 81-120), which contains a PDZ-domain binding motif (aa 117-120) that interacts with the Maguk protein, PALS1, which recruits PALS1-associated tight junction (PATJ) protein into the CRB3A complex. Both PALS1 and PATJ play a critical role in epithelial cell polarity and tight junction formation. CRB3A is shown to control and properly localize RhoA signaling that is required for CRB3A-dependent reorganization of the actin cytoskeleton. (Ref.: Loie, E., et al (2015). Mol. Cell Biol. 35(19); 3423-3435).

Specificity

Clone 1E6 is a rat monoclonal antibody that detects protein crumbs homolog 3 in human cells. it targets an epitope with in 15 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Protein crumbs homolog 3 (CRB3).

Application

Anti-CRB3A, clone 1E6, Cat. No. MABT1366, is a rat monoclonal antibody that detects Protein crumbs homolog 3 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Cell Structure
Western Blotting Analysis: 2 µg/mL from a representative lot detected CRB3A in Caco2 cell lysates over-expressing Crumbs 3A (positive) versus lysates from Caco2 cells silenced with siRNA (negative).

Western Blotting Analysis: A representative lot detected CRB3A in Western Blotting applications (Loie, E., et. al. (2015). Mol Cell Biol. 35(19):3423-35).

Quality

Evaluated by Immunohistochemistry (Paraffin) in human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected CRB3A in human kidney tissue sections.

Target description

~35 kDa observed; 12.85 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Mangeol et al.
eLife, 11 (2022-11-08)
Epithelial tissues acquire their integrity and function through the apico-basal polarization of their constituent cells. Proteins of the PAR and Crumbs complexes are pivotal to epithelial polarization, but the mechanistic understanding of polarization is challenging to reach, largely because numerous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service