Skip to Content
Merck
All Photos(2)

Documents

MABS434

Sigma-Aldrich

Anti-PCSK9 Antibody, clone 9B3.1

culture supernatant, clone 9B3.1, from mouse

Synonym(s):

Proprotein convertase subtilisin/kexin type 9, NARC-1, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, Subtilisin/kexin-like protease PC9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

9B3.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PCSK9(255738)

General description

Proprotein convertase subtilisin/kexin type 9 (UniProt Q8NBP7; also known as NARC-1, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, Subtilisin/kexin-like protease PC9) is encoded by the PCSK9 (also known as HCHOLA3, LDLCQ1, NARC1, PSEC0052) gene (Gene ID 255738) in human. PCSK9 is a proprotein convertase expressed in the hepatocyte and to a lesser extent also in the intestine and kidney. PCSK9 is produced as a precusor protein with a signal peptide (a.a.1-30) and a propeptide sequence (a.a. 31-152) that undergo posttranslational autocleavage to yield the mature PCSK9 protein (a.a. 153-692). PCSK9 utilizes its proteolytic activity only in autocleavage during protein maturation, a process that is necessary for its secretion. Once secreted into the plasma, PCSK9 binds LDL receptor (LDL-R), causing LDL-R endocytosis and lysosomal degradation, effectively reducing cell surface LDL-R population. Both PCSK9 and LDL-R are transcribed by sterol-responsive element-binding protein 2 (SREBP2) in response to lowered cellular cholesterol level, allowing enhanced, but not unfettered cellular cholesterol uptake by LDL-R. PCSK9 also binds very low density lipoprotein (VLDL) receptor and apolipoprotein E receptor 2, and thereby interferes with uptake of circulating VLDL particles and apolipoproteins.

Specificity

Clone 9B3.1 detects the precursor and mature forms of human PCSK9 (NARC-1) by targeting an epitope within the N-terminal region of the mature sequence.

Immunogen

GST-tagged recombinant human mature NARC-1 (PCSK9) N-terminal fragment.

Application

Detect PCSK9 using this mouse monoclonal Anti-PCSK9, clone 9B3.1 Antibody, Cat. No. MABS434, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected NARC-1 (PCSK9) in human colon cancer, normal kidney and prostate tissue sections.

Western Blotting Analysis: A 1:500 dilution from a representative lot detected NARC-1 (PCSK9) in 10 µg of human colon tumor tissue lysate.
Research Category
Signaling

Quality

Evaluated by Western Blotting in SW480 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this hybridoma culture supernatant detected NARC-1 (PCSK9) in 10 µg of SW480 human colon adenocarcinoma cell lysate.

Target description

~75/60 kDa observed. 74.29 kDa (prepro-form; a.a. 1-692), 71.05 kDa (pro-form; a.a. 31-692), and 57.29 kDa (mature form; a.a. 153-692) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse IgG2a hybridoma culture supernatant without azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service