MABS2309
Anti-p66shc Antibody, clone BG8
Synonym(s):
SHC-transforming protein 1;SHC-transforming protein 3;SHC-transforming protein A;Src homology 2 domain-containing-transforming protein C1;SH2 domain protein C1
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
BG8, monoclonal
mol wt
calculated mol wt 62.82 kDa
observed mol wt ~62 kDa
purified by
using protein G
species reactivity
mouse
species reactivity (predicted by homology)
human
packaging
antibody small pack of 100 μL
technique(s)
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable
isotype
IgG1κ
epitope sequence
N-terminal
Protein ID accession no.
UniProt accession no.
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... SHC1(6464)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in NIH3T3 cell lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected SHC-transforming protein 1 in NIH3T3 cell lysate.
Tested Applications
Immunofluorescence Analysis: A representative lot detected p66shc 1 in Immunofluorescence application (Orsini, F., et al. (2004). J Biol Chem. 279(24): 25689-95).
Immunocytochemistry Analysis: A representative lot detected p66shc in Immunocytochemistry application (Orsini, F., et al. (2004). J Biol Chem. 279(24): 25689-95).
Western Blotting Analysis: A representative lot detected p66shc in Western Blotting application (Orsini, F., et al. (2004). J Biol Chem. 279(24): 25689-95); Su, K.G., et al. (2012). Eur J Neurosci. 35(4):562-71).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service