Skip to Content
Merck
All Photos(1)

Key Documents

MABS2292

Sigma-Aldrich

Anti-SMCHD1 Antibody, clone 2B8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B8 (#8), monoclonal

mol wt

calculated mol wt 225.65 kDa
observed mol wt ~225 kDa

species reactivity

mouse

species reactivity (predicted by homology)

human

packaging

antibody small pack of 100 μL

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Structural maintenance of chromosomes flexible hinge domain-containing protein 1 (UniProt: Q6P5D8; also known as SMC hinge domain-containing protein 1) is encoded by the Smchd1 (also known as Kiaa0650) gene (Gene ID: 74355) in murine species. SMCHD1 is a homodimeric, non-canonical member of the structural maintenance of chromosomes (SMC) protein family that plays a key role in epigenetic silencing by regulating chromatin architecture. It is shown to be essential for X chromosome inactivation. It is recruited to inactivated chromosome X by Xist RNA and acts by mediating the merge of chromatin compartments. It is reported to localize to the inactive X via the XistHnrnpK-polycomb repressive complex 1 pathway. In addition to its role in X inactivation, it also plays a role in the expression of several autosomal gene clusters that are subject to monoallelic expression. It promotes heterochromatin formation in both autosomes and chromosome X, probably by mediating the merge of chromatin compartments. SMCHD1 also plays a role in DNA repair. It localizes to sites of DNA double-strand breaks in response to DNA damage to promote the repair of DNA double-strand breaks. It acts by promoting non-homologous end joining and inhibiting homologous recombination repair. Heterozygous mutations in Smchd1 gene have been linked to facioscapulohumeral muscular dystrophy 2, an inherited muscular weakness syndrome. (Ref.: Jansz, N., et al. (2018). Cell Rep. 25(7); 1912-1923; Mould, AW., et al. (2013). Epigenetics Chromatin 6(1); 19; Blewitt, ME., et al. (2008). Nat. Genet. 40(5); 663-669).

Specificity

Clone 2B8 is a rat monoclonal antibody that detects SMC hinge domain-containing protein 1 (SMCHD1). It targets an epitope within the ATPase activity domain in the N-terminal half.

Immunogen

Recombinant fragment corresponding to 592 amino acids from the ATPase activity domain from the N-terminal half of murine Structural maintenance of chromosomes flexible hinge domain-containing protein 1 (SMCHD1).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from wild-type Mouse neuronal stem cells.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected Structural maintenance of chromosomes flexible hinge domain-containing protein 1 (SMCHD1) in lysate from wild-type Mouse neuronal stem cells, but not in Smchd1 null cells.

Tested Applications

Western Blotting Analysis: A representative lot detected SMCHD1 in Western Blotting applications (Jansz, N., et. al. (2018). Cell Rep. 25(7):1912-1923).

Immunofluorescence Analysis: A representative lot detected SMCHD1 in Immunofluorescence applications (Jansz, N., et. al. (2018). Cell Rep. 25(7):1912-1923).

Immunoprecipitation Analysis: A representative lot immunoprecipitated SMCHD1 in Immunoprecipitation applications (Jansz, N., et. al. (2018). Cell Rep. 25(7):1912-1923).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SMCHD1, clone 2B8, Cat. No. MABS2292, is a rat monoclonal antibody that detects Structural maintenance of chromosomes flexible hinge domain-containing protein 1 (SMCHD1) and is tested in Immunofluorescence, Immunoprecipitation, and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service