Skip to Content
Merck
All Photos(1)

Key Documents

MABS1917

Sigma-Aldrich

Anti-RIP140 Antibody, clone 6D7

clone 6D7, from mouse

Synonym(s):

Nuclear receptor-interacting protein 1, Nuclear factor RIP140, Receptor-interacting protein 140

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D7, monoclonal

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μg

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... NRIP1(8204)

General description

Nuclear receptor-interacting protein 1 (UniProt: P48552; also known as Nuclear factor RIP140, Receptor-interacting protein 140) is encoded by the NRIP1 gene (Gene ID: 8204) in human. RIP140 is a nuclear protein that is localized to discrete foci and redistributes to larger nuclear domains upon binding to ligand-bound NR3C1. It is expressed in liver in a circadian manner. RIP140 contains 9 Leu-Xaa-Xaa-Leu-Leu (LXXLL) motifs, which have different affinities for nuclear receptors. The C-terminal LTKTNPILYYMLQK motif is required for ligand-dependent interaction with retinoic acid (RA) receptor-alpha (RARA) and retinoid X receptor beta (RXRB) homodimers and heterodimers, for the corepressor activity, and for the formation of an HDAC3 complex with RARA/RXRB. It also contains at least four autonomous repression domains (RD1-4). RD1 functions via a histone deacetylase (HDAC)-independent mechanism, whereas RD2, RD3 and RD4 can function by HDAC-dependent or independent mechanisms, depending on cell type. RIP140 Acetylation regulates its nuclear translocation and co-repressive activity of RIP140 is regulated by its acetylation, which is shown to abolish its interaction with C-terminal-binding protein 1 (CTBP1). Several glucocorticoid responses are shown to be deregulated by RIP140 possibly via an interaction between the glucocorticoid receptor (GR), which prevents binding of true co-activator with GR. (Ref.: Subramaniam N, et al. (1999). J. Biol. Chem. 274(25):18121-7).

Specificity

Clone 6D7 detects Nuclear receptor-interacting protein 1 (RIP140) in human, mouse, and rat cells. It targets an epitope with in 178 amino acids from the N-terminal half.

Immunogen

GST-tagged recombinant fragment corresponding to 178 amino acids from the N-terminal half of human Nuclear receptor-interacting protein 1 (RIP140).

Application

Anti-RIP140, clone 6D7, Cat. No. MABS1917, is a mouse monocloanl antibody that detects RIP140 and has been tested for use in Chromatin Immunoprecipitation (ChIP) and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected RIP140 in Western Blotting applications (Kiskinis, E., et. al. (2007). EMBO J. 26(23):4831-40; Hallberg, M., et. al. (2008). Mol Cell Biol. 28(22):6785-95).

Chromatin Immunoprecipitation Analysis: A representative lot detected RIP140 in Chromatin Immunoprecitpiation applications (Hallberg, M., et. al. (2008). Mol Cell Biol. 28(22):6785-95).

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected RIP140 in 10 µg of MCF7 cell lysate.

Target description

~160 kDa observed; 126.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara A Harper et al.
Experimental gerontology, 143, 111111-111111 (2020-10-18)
To evaluate the safety and feasibility of combining exercise (EX) and resveratrol to treat older adults with physical function limitations. Three-arm, two-site pilot randomized, controlled trial (RCT) for community-dwelling adults (N = 60), 71.8 ± 6.3 years of age with functional limitations. Participants were randomized
Mijeong Kim et al.
Nature communications, 15(1), 1285-1285 (2024-02-13)
During human pregnancy, extravillous trophoblasts play crucial roles in placental invasion into the maternal decidua and spiral artery remodeling. However, regulatory factors and their action mechanisms modulating human extravillous trophoblast specification have been unknown. By analyzing dynamic changes in transcriptome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service