Skip to Content
Merck
All Photos(1)

Key Documents

MABS1173

Sigma-Aldrich

Anti-E3 ubiquitin-protein ligase CBL-C Antibody, clone 10F4.2

clone 10F4.2, from mouse

Synonym(s):

E3 ubiquitin-protein ligase CBL-C, RING finger protein 57, SH3-binding protein CBL-3, SH3-binding protein CBL-C, Signal transduction protein CBL-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10F4.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CBLC(23624)

General description

E3 ubiquitin-protein ligase CBL-C (UniProt Q9ULV8; also known as RING finger protein 57, SH3-binding protein CBL-3, SH3-binding protein CBL-C, Signal transduction protein CBL-C) is encoded by the CBLC (also known as CBL3, RNF57) gene (Gene ID 23624) in human. CBLC belongs to the CBL family of E3 ubiquitin ligases, members of which also include CBL (a.k.a. c-CBL) and CBL-B. Similar to CBL and CBL-B, CBL-C binds SH3 (Src homology 3) domain-containing proto-oncogenic tyrosine kinases such as EGFR, Lyn, Crk, Src and Ret. CBL-C is recruited to EGFR upon EGF stimulation, leading to CBL-C phosphorylation by EGFR and subsequent downstream MAP kinase signalling. In addition to mediating oncoprotein Ret ubiquitination for proteasomal degradation, CBL-C is shown to form a complex with HIC5 (Hydrogen peroxide-inducible clone 5) and the heat shock protein HSP27 that are necessary for the ubiquitination of NOX4. Unlike the other CBL family members, CBL-C expression appears to be restricted to epithelial tissues. Transgenic expression of CBL-C in mouse mammary gland causes impairment of cell proliferation, suggesting that CBL-C might have a growth-inhibitory role. CBL-C contains an N-terminal Cbl-type phosphotyrosine-binding (Cbl-PTB) domain (a.a. 7-321) that mediates SH3 domain interaction and a C-terminal RET-interacting region (a.a. 351-474) harboring the catalytic ring finger domain (a.a. 351-390) that mediates interaction with E2 ubiquitin-conjugating enzyme. The PTB domain, also known as tyrosine kinase-binding (TKB) domain, is subdivided into a four-helix bundle or 4H domain (a.a. 7-145), a calcium-binding EF hand-like domain (a.a. 146-218), and a SH2-like domain (a.a. 219-321).

Specificity

Clone 10F4.2 targets an epitope in the RET-interacting region present in both spliced isoforms of human CBL-C reported by UniProt (Q9ULV8).

Immunogen

Epitope: RET-interacting region.
GST-tagged recombinant fragment from the RET-interacting region of human CBL-C.

Application

Research Category
Signaling
Research Sub Category
Ubiquitin & Ubiquitin Metabolism
This Anti-E3 ubiquitin-protein ligase CBL-C Antibody, clone 10F4.2 is validated for use in Western Blotting for the detection of E3 ubiquitin-protein ligase CBL-C.

Quality

Evaluated by Western Blotting in HT-29 human colon cancer cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected CBL-C in 10 µg of HT-29 human colon cancer cell lysate.

Target description

~50 kDa observed. 52.46 kDa (isoform 1; Long) and 47.42 kDa (isoform 2; Short) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service