Skip to Content
Merck
All Photos(4)

Documents

MABS1139

Sigma-Aldrich

Anti-SUMO2/3 Antibody, clone 3H12

clone 3H12, from rat

Synonym(s):

Small ubiquitin-related modifier 2, HSMT3, Sentrin-2, SMT3 homolog 2, Smt3B, SUMO-2, Ubiquitin-like protein SMT3B, Small ubiquitin-related modifier 3, SMT3 homolog 1, Smt3A, SUMO-3, Ubiquitin-like protein SMT3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rat

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

3H12, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SUMO2(6613)

General description

Small ubiquitin-related modifier 2 (UniProt P61956; a.k.a. HSMT3, Sentrin-2, SMT3 homolog 2, Smt3B, SUMO-2, Ubiquitin-like protein SMT3B) and 3 (UniProt P55854; a.k.a. SMT3 homolog 1, Smt3A, SUMO-3, Ubiquitin-like protein SMT3A) are encoded by the SUMO2 (a.k.a. SMT3B, SMT3H2; Gene ID 6613) and SUMO3 (a.k.a. SMT3A, SMT3H1; Gene ID 6612) genes in human. SUMOylation, protein posttranslation modification by small ubiquitin-like modifier (SUMO), is a signaling event in many cellular processes. SUMO proteins are translated as immature precursors and subsequently converted to their mature forms through the activity of sentrin/SUMO-specific proteases (SENPs). As is the case with ubiquitination, SUMOylation is a reversible process. SUMO E1 activating enzyme, E2 conjugating enzyme, and E3 ligase mediate SUMOylation of substrate proteins, while SENPs are responsible for the deSUMOylation. SUMOylation usually occurs at lysine residues in the consensus KxD/E motif, although not all such lysines become SUMOylated and SUMOylation can also occur on lysine residues outside of this motif. SUMO2 and 3 share 97% identity at the amino acid level, while SUMO1 shares approximately 50% and SUMO4 shares about 87% identity with SUMO2/3. SUMO4 is the only SUMO member whose ctivation and conjugation has not been demonstrated. In addition to difference in their target substrates, SUMO2/3 can be SUMOylated and form chains, whereas SUMO1 cannot and may serve as chain terminator.

Specificity

Clone 3H12 detects SUMO2/3 and SUMO2/3-modified (SUMOylated) proteins. Clone 3H12 reacts with SUMO2/3, but not SUMO1 (Uchimura, Y., et al. (2006). J. Biol. Chem. 281(32):23180-23190).
Predicted to react with a broad-range of mammalian species based on high sequence homology.

Immunogen

GST-tagged recombinant full-length human SUMO3 (Uchimura, Y., et al. (2006). J. Biol. Chem. 281(32):23180-23190).

Application

Detect SUMO2/3 and SUMO2/3-modified (SUMOylated) proteins using this rat monoclonal Anti-SUMO2/3 Antibody, clone 3H12, Cat. No. MABS1139, validated for use in Immunocytochemistry and Western blotting.
Immunocytochemistry Analysis: Undiluted hybridoma culture supernatant from a representative lot detected SUMO2/3 immunoreactivity in 4% paraformaldehyde-fixed, 0.3% Triton X-100-permeabilized HeLa cells.

Immunocytochemistry Analysis: 10 µg/mL of purified clone 3H12 detected SUMO2/3 immunoreactivity in primary mouse neural progenitor cells (Courtesy of Taro Tachibana, Ph.D., Osaka City University, Japan).

Western Blotting Analysis: 1 µg/mL of purified clone 3H12 detected SUMOylated proteins in HeLa cell lysate (Courtesy of Taro Tachibana, Ph.D., Osaka City University, Japan).

Immunocytochemistry Analysis: A representative lot detected SUMO-2/3 nuclear immunoreactive foci co-localized with those of MCAF1 and SUMO1 and enriched in heterochromatin proteins by indirect immunofluorescent staining of exponentially growing C-33A cells fixed with 4% paraformaldehyde and permeabilized by 0.2% Triton X-100. Trasfecting C-33A cells with SUMO2/3 shRNA or Discosoma sp. red fluorescent protein (DsRed) fusion containg SUMO2/3-interacting sequence (MCAF1 a.a. 965-975) abolished nuclear staining by clone 3H12 (Uchimura, Y., et al. (2006). J. Biol. Chem. 281(32):23180-23190).
Research Category
Signaling

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this hybridoma culture supernatant detected SUMO2/3-modified (SUMOylated) proteins in 10 µg of HeLa cell lysate.

Target description

Variable depending on the degree of SUMOylation and the size(s) of the SUMOylated protein(s). 10.61/10.52 kDa (SUMO2/SUMO3; propeptide removed) calculated.

Physical form

Format: Unpurified
Rat IgG2a hybridoma culture supernatant with 0.05% sodium azide.
Unpurified.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service