Skip to Content
Merck
All Photos(2)

Key Documents

MABS107-I

Sigma-Aldrich

Anti-Ubiquitin K11 linkage Antibody, clone 2A3/2E6

clone 2A3/2E6, from rabbit

Synonym(s):

Ubiquitin K11 linkage, Lysine 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A3/2E6, monoclonal

species reactivity

human

species reactivity (predicted by homology)

all (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UBAP2(55833)

General description

Recently, the lysine11 (K11) linkage of ubiquitin has been shown to be specifically targeted by the Anaphase-Promoting Complex (APC) E3 ubiquitin ligase for catalyzed ubiquitination for mitosis. Antibodies specific to the K11-linkage have shown that the formation of the K11 chains are greatly increased in mitotic human cells in the presence of APC substrate degradation. K11-linked ubiquitin chains seem to have essential regulatory control over mitotic protein degradation.

Immunogen

Recombinant protein corresponding to all in Ubiquitin K11 linkage.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Ubiquitin K11 linkage in A431 and HeLa cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling
This Anti-Ubiquitin K11 linkage Antibody, clone 2A3/2E6 is validated for use in western blotting & ICC for the detection of Ubiquitin K11 linkage.

Quality

Evaluated by Western Blotting in K11 recombinant protein.

Western Blotting Analysis: A 1:250 to 1:1,000 dilution of this antibody detected Ubiquitin K11 linkage in 10 µg of K11 recombinant protein.

Target description

Varies

Physical form

Format: Purified
Protein A purified
Purified rabbit monoclonal IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingwei Min et al.
Molecular biology of the cell, 26(24), 4325-4332 (2015-10-09)
The ubiquitin proteasome system (UPS) directs programmed destruction of key cellular regulators via posttranslational modification of its targets with polyubiquitin chains. These commonly contain Lys-48 (K48)-directed ubiquitin linkages, but chains containing atypical Lys-11 (K11) linkages also target substrates to the
Michael E French et al.
The Journal of biological chemistry, 292(25), 10398-10413 (2017-05-04)
Homologous to E6AP C-terminal (HECT) ubiquitin (Ub) ligases (E3s) are a large class of enzymes that bind to their substrates and catalyze ubiquitination through the formation of a Ub thioester intermediate. The mechanisms by which these E3s assemble polyubiquitin chains
Rahul S Samant et al.
Nature, 563(7731), 407-411 (2018-11-16)
Protein misfolding is linked to a wide array of human disorders, including Alzheimer's disease, Parkinson's disease and type II diabetes1,2. Protective cellular protein quality control (PQC) mechanisms have evolved to selectively recognize misfolded proteins and limit their toxic effects3-9, thus
Swarna L Vijayaraj et al.
Nature communications, 12(1), 2713-2713 (2021-05-13)
Interleukin-1β (IL-1β) is activated by inflammasome-associated caspase-1 in rare autoinflammatory conditions and in a variety of other inflammatory diseases. Therefore, IL-1β activity must be fine-tuned to enable anti-microbial responses whilst limiting collateral damage. Here, we show that precursor IL-1β is
Animesh Dhara et al.
mSphere, 1(3) (2016-06-25)
The contribution of ubiquitin-mediated mechanisms in the regulation of the Toxoplasma gondii cell cycle has remained largely unexplored. Here, we describe the functional characterization of a T. gondii deubiquitinase (TGGT1_258780) of the ovarian-tumor domain-containing (OTU) family, which, based on its structural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service