Skip to Content
Merck
All Photos(1)

Key Documents

MABN782

Sigma-Aldrich

Anti-Amyloid-like protein 2, clone 5C9.3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

Quality Level

clone

5C9.3
monoclonal

purified by

using protein G

species reactivity

human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

General description

GST-tagged recombinant protein corresponding to human Amyloid-like protein 2.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Amyloid-like protein 2 in human kidney and human cerebral cortex tissue.

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Amyloid-like protein 2 in 10 µg of SH-SY5Y cell lysate.

Physical properties

~87 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service