MABN2472
ANTI-TAU (MAPT) Antibody
mouse monoclonal, 2A1-2E1
Synonym(s):
Microtubule-associated protein tau, Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau
About This Item
Recommended Products
Product Name
Anti-Tau Antibody, clone 2A1-2E1, clone 2A1-2E1, from mouse
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2A1-2E1, monoclonal
species reactivity
mouse, human
packaging
antibody small pack of 25 μg
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... MAPT(4137)
mouse ... Mapt(17762)
General description
Specificity
Immunogen
Application
Neuroscience
Immunohistochemistry Analysis: A representative lot detected Tau in Immunohistochemistry applications (Ishigaki, S., et. al. (2017). Cell Rep. 18(5):1118-1131).
Quality
Western Blotting Analysis: 2 µg/mL of this antibody detected Tau in human Tau-441 recombinant protein lysates.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service