Skip to Content
Merck
All Photos(2)

Key Documents

MABN1846

Sigma-Aldrich

Anti-Apelin Receptor/APJ, clone 3C3-7 Antibody

clone 3C3-7, from mouse

Synonym(s):

Apelin receptor, Angiotensin receptor-like 1, G-protein coupled receptor APJ, G-protein coupled receptor HG11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C3-7, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... APLNR(187)

General description

Apelin receptor (UniProt P35414; also known as Angiotensin receptor-like 1, G-protein coupled receptor APJ, G-protein coupled receptor HG11) is encoded by the APLNR (also known as AGTRL1, APJ) gene (Gene ID 187) in human. Apelin is an angiotensin converting enzyme 2 (ACE2) substrate that functions as an inotropic and cardiovascular protective peptide. Apelin exerts its physiological effects via its receptor APJ, a seven-transmembrane G protein-coupled receptor that shares significant homology with the Ang II type 1 receptor (AT1R). Apelin/APJ signaling plays an important role in a wide range of pathophysiological effects in the cardiovascular system. Apelin levels are reduced in patients with essential hypertension independent of left ventricular systolic and diastolic dysfunction. Treatment with Apelin has been shown to lower systolic blood pressure levels in hypertensive rats via an NO-dependent mechanism. APJ is a 7-transmembrane (a.a. 27-51, 67-91, 101-125, 145-166, 201-221, 245-271, 285-308) GPCR, having a short N-terminal extracellular portion (a.a. 1-26) and a long C-terminal cytomplasmic tail (a.a. 309-380).

Specificity

Clone 3C3-7 detects human Apelin receptor/APJ.

Immunogen

Recombinant full-length human Apelin receptor/APJ.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Apelin receptor/APJ in human prostate cancer tissue, as well as normal cerebral cortex and pancreas tissue sections.
Research Category
Neuroscience
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling
This Anti-Apelin Receptor, clone 3C3-7 Antibody is validated for use in Immunohistochemistry (Paraffin), Western Blotting for the detection of APJ.

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Apelin receptor/APJ in 10 µg of MCF7 cell lysate.

Target description

~42 kDa 42.66 kDa calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sahil Gulati et al.
Nature communications, 9(1), 1996-1996 (2018-05-20)
G protein-coupled receptors (GPCRs) activate heterotrimeric G proteins by mediating a GDP to GTP exchange in the Gα subunit. This leads to dissociation of the heterotrimer into Gα-GTP and Gβγ dimer. The Gα-GTP and Gβγ dimer each regulate a variety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service