Skip to Content
Merck
All Photos(1)

Key Documents

MABF934

Sigma-Aldrich

Anti-phospho-SAMHD1 (Thr592) Antibody, clone T592P

clone T592P, from mouse

Synonym(s):

Deoxynucleoside triphosphate triphosphohydrolase SAMHD1, dNTPase, Dendritic cell-derived IFNG-induced protein, DCIP, Monocyte protein 5, MOP-5, SAM domain and HD domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

T592P, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pThr592)

Gene Information

human ... SAMHD1(25939)

General description

Deoxynucleoside triphosphate triphosphohydrolase SAMHD1 (UniProt: Q9Y3Z3; also known as dNTPase, Dendritic cell-derived IFNG-induced protein, DCIP, Monocyte protein 5, MOP-5, SAM domain and HD domain-containing protein 1) is encoded by the SAMHD1 (also known as MPOS) gene (Gene ID: 25939) in human. SAMHD1 is a nuclear protein that functions as a host restriction nuclease that blocks early-stage virus replication in dendritic and other myeloid cells. It suppresses LINE-1 retrotransposon activity and may function by reducing the cellular dNTP to such low levels that prevents retroviral reverse transcription. SAMHD1 is also reported to play a role in mediating pro-inflammatory responses to TNF-alpha signaling. It is expressed in heart, skeletal muscle, spleen, liver, small intestine, placenta, lung and peripheral blood leukocytes, however, no expression is seen in brain and thymus tissue. Its expression is shown to be upregulated in TNF-alpha treated fibroblasts. SAMHD1 is allosterically stimulated by dGTP, which binds in a cleft at the interface of the homodimer and promotes the formation of highly active homotetramers. Four isoforms of SAMHD1 have been described that are produced by alternate splicing. Mutations in SAMHD1 gene are reported to be involved in the pathogenesis of Chilblain lupus 2, a cutaneous form of lupus erythematosus and in Aicardi-Goutieres syndrome 5 that is characterized by cerebral atrophy, leukoencephalopathy, intracranial calcifications, chronic cerebrospinal fluid (CSF) lymphocytosis.

Specificity

Clone T592P detects Deoxynucleoside triphosphate triphosphohydrolase SAMHD1 in human cells. It targets an epitope with in 11 amino acids surrounding the phosphorylated Thr592 from the C-terminal region.

Immunogen

A linear peptide corresponding to 11 amino acids surrounding phosphoryalted Thr592 from the C-terminal region of huamn SAMHD1.

Application

Anti-phospho-SAMHD1 (Thr592), clone T592P, Cat. No. MABF934 is a mouse monoclonal antibody that detects SAMHD1 and has been tested for use in Flow Cytometry and Western Blotting.
Flow Cytometry Analysis: A representative lot detected phospho-SAMHD1 (Thr592) in Flow Cytometry applications (Coiras, M., et. al. (2016). Cell Rep. 14(9):2100-7).

Western Blotting Analysis: A representative lot detected phospho-SAMHD1 (Thr592) in Western Blotting applications (Cribier, A., et. al. (2013). Cell Rep. 3(4):1036-43; Calantone, N., et. al. (2014). Immunity. 41(3):493-502; Coiras, M., et. al. (2016). Cell Rep. 14(9):2100-7).
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in CD4+ T lymphocytes (activated with PMA/IL-2) lysates.

Western Blotting Analysis: A 1:500 dilution of this antibody detected phospho-SAMHD1 (Thr592) in CD4+ T lymphocytes (activated with PMA/IL-2) lysates.

Target description

~72 kDa observed; 72.20 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service