Skip to Content
Merck
All Photos(1)

Key Documents

MABF3081

Anti-SLAMF6/NTB-A Antibody, clone NT-7

new

Synonym(s):

Activating NK receptor, CD352, Lymphocyte antigen 108, NK-T-B-antigen (NTB-A), SLAM family member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

NT-7, monoclonal

mol wt

calculated mol wt 37 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
flow cytometry: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... SLAMF6(114836)

Specificity

Clone NT-7 is a mouse monoclonal antibody that specifically detects SLAMF6/NTB-A. It targets an epitope within the extracellular domain.

Immunogen

Recombinant fragment from extracellular domain of human SLAMF6/NTB-A.

Application

Quality Control Testing

Evaluated by Flow Cytometry in Human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis: 1.0ug of this antibody detected SLAMF6/NTB-A in one million Human peripheral blood mononuclear cells (PBMC).

Tested Applications

ELISA: A representative lot detected NTB-A/CD352 in ELISA application (Stark, S., et al. (2005). J Immunol Methods. 296(1-2):149-58).

Flow Cytometry Analysis: A representative lot detected NTB-A/CD352 in Flow Cytometry application (Flaig, R.M., et al. (2004). J Immunol. 172(11):6524-7; Messmer, B., et al. (2006). J Immunol. 176(8):4646-50).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

SLAM family member 6 (UniProt: Q96DU3; also known as Activating NK receptor, NK-T-B-antigen (NTB-A), CD352) is encoded by the SLAMF6 (also known as KALI) gene (Gene ID: 114836) in human. SLAMF6 belongs to the 7-member signaling lymphocytic activation molecule family (SLAMF) of receptors broadly expressed in hematopoietic cells. SLAM receptors triggered by homo or heterotypic cell-cell interactions are modulating the activation and differentiation of a wide variety of immune cells and thus are involved in the regulation and interconnection of both innate and adaptive immune response. SLAMF members carry one or more copies of immunoreceptor tyrosine-based switch motif or ITSM (T-X-Y-X-X-[VI]) in their cytoplasmic tails. Upon phosphorylation on the tyrosine residue, ITSM mediates the recruitment of SH2 domain-containing signaling molecules such as SLAM-associated protein (SAP), including SAP in T-, NK, and NK T-cells, EAT-2A and EAT-2B (murine) in NK cells and APCs. SLAMF6/NTB-A is predominantly localized to the cell membrane and is a single-pass type I membrane protein and is synthesized with a signal peptide sequence (aa 1-21), the removal of which yields the mature SLAMF6 with an extracellular domain (aa 22-226), a transmembrane domain (aa 227-247) and, a cytoplasmic domain (aa 248-331). Its expression is primarily observed in cells of the immune system. SLAMF6/NTB-A is a receptor on the surfaces of human NK, T, and B cells, mediating a signal whose malfunction may be involved in X-linked lymphoproliferative disease. Studies show that NTB-A is its own ligand and homophilic interaction of NTB-A enhances NK cell cytotoxicity and influences NK cell proliferation and IFN- secretion. It is also suggested that NTB-A is an interlymphocyte signaling molecule, which serves to orchestrate the activities of immune cells. Dysregulation of SLAMF6 signaling is associated with autoimmune conditions such as systemic lupus erythematosus (SLE), where it can contribute to abnormal immune cell activation. (Ref.: Flaig, R.M., et al. (2004). J Immunol. 172(11):6524-7; Stark, S., et al. (2005). J Immunol Methods. 296(1-2):149-58; Messmer, B., et al. (2006). J Immunol. 176(8):4646-50).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service