Skip to Content
Merck
All Photos(1)

Key Documents

MABF116

Sigma-Aldrich

Anti-CD3 gamma delta (HMT3-2) Antibody, clone Ham25-1157

clone Ham25-1157, from hamster

Synonym(s):

T-cell surface glycoprotein CD3 gamma chain, T-cell receptor T3 gamma chain, CD3g, HMT3-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

hamster

antibody form

purified antibody

antibody product type

primary antibodies

clone

Ham25-1157, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

flow cytometry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD3G(917)

General description

The CD3 complex is composed of six polypeptides with usually four different transmembrane CD3 chains: gamma, delta, epsilon and zeta. CD3 is first detectable in early thymocytes and one of the earliest signs of commitment to the T cell lineage. In cortical thymocytes, during early stages of maturation, the CD3 antigen is predominantly present in the cell cytoplasm. In medullary thymocytes, it is detected on the cell surface. The majority of T cell neoplasms also expresses CD3, but it is absent from non-T-cell lymphoid malignancies.

Specificity

This antibody recognizes CD3 gamma delta.

Immunogen

Purified protein corresponding to mouse CD3 gamma delta.

Application

Anti-CD3 gamma delta (HMT3-2) Antibody, clone Ham25-1157 detects level of CD3 gamma delta (HMT3-2) & has been published & validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Flow Cytometry in Jurkat cells.

Flow Cytometry Analysis: 2.5 µg/mL of this antibody detected CD3 gamma delta in 1X10E5 Jurkat cells.

Target description

21 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified armenian hamster monoclonal IgG2bλ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service