Skip to Content
Merck
All Photos(2)

Key Documents

MABF1105

Sigma-Aldrich

Anti-DAG Kinase Alpha Antibody, clone 10D6.1

clone 10D6.1, from mouse

Synonym(s):

Diacylglycerol kinase alpha, 80 kDa diacylglycerol kinase, Diglyceride kinase alpha, DGK-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10D6.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... DGKA(1606)

General description

Diacylglycerol kinase alpha (UniProt: P23743; also known as EC:2.7.1.107, DAG kinase alpha, 80 kDa diacylglycerol kinase, Diglyceride kinase alpha, DGK-alpha) is encoded by the DGKA (also known as DAGK, DAGK1) gene (Gene ID: 1606) in human. Diacylglycerol kinase is a lipid kinase that converts diacylglycerol to phosphatidic acid, and regulates the activity of many enzymes including protein kinase C, phosphatidylinositol 4-phosphate 5-kinase, and mTOR. Thus far ten mammalian subtypes of DAG kinase have been reported. All DAG kinase with the exception of beta isoform are localized in cytoplasm in several cells, but some translocate to the plasma membrane upon activation. DAG kinase acts as a modulator, which competes with protein kinase C for diacylglycerol in intracellular signaling pathways. DAG kinase also plays a role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Most DAG kinases, with the exception of theta, contain two cysteine-rich regions (C1A and C1B domains) in the regulatory domain of the N-terminal half of the molecule. In DAK kinase alpha the C1 domain closest to the catalytic domain has an extended region of fifteen amino acids that is not present in C1 domains of other DAG kinases. DAG kinase alpha can be activated by the phosphorylation of Tyr335, which occurs during T cell activation or following exposure of cells to hepatocyte growth factor.

Specificity

Clone 10D6.1 detects DAG kinase alpha in human. It targets an epitope within the first 110 amino acids from the N-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to the first 110 amino acids of human DAG kinase alpha.

Application

Anti-DAG Kinase Alpha, clone 10D6.1, Cat. No. MABF1105, is a mouse monoclonal antibody that detects DAG kinase alpha. It has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected DAG Kinase Alpha in human tonsil and human breast cancer tissue.
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected DAG Kinase Alpha in 10 µg of Jurkat cell lysate.

Target description

~83 kDa observed; 82.63 kDa calculaed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service